Assay Details
Target Gene Details
Entrez Gene ID: | 171586 |
Gene Name: | abhydrolase domain containing 3 |
Gene Aliases: |
LABH3 |
Location: |
Chr.18:21650897-21704805 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABHD3 | NM_001308256.1 | 4 | 740 | NP_001295185.1 |
NM_001308257.1 | 2 | 628 | NP_001295186.1 | |
NM_138340.4 | 6 | 899 | NP_612213.2 | |
XM_011525847.2 | 6 | 872 | XP_011524149.1 | |
XM_017025571.1 | 6 | 1315 | XP_016881060.1 | |
XM_017025572.1 | 5 | 1143 | XP_016881061.1 | |
XM_017025573.1 | 7 | 1124 | XP_016881062.1 | |
XM_017025574.1 | 5 | 1067 | XP_016881063.1 | |
AF007152.1 | 6 | 858 | AAC19155.1 | |
AK024880.1 | 6 | 773 | ||
AK024992.1 | 2 | 108 | ||
AK298728.1 | 6 | 1238 | ||
AV654056.1 | ||||
BC021196.2 | 6 | 859 | AAH21196.1 | |
BG547174.1 | 2 | 240 | ||
BI918326.1 | 4 | 692 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv29974 | Chr.18:21593909 - 21668731 on Build GRCh38 | Loss | SNRPD1 ABHD3 ESCO1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |