Assay Details
Target Gene Details
Entrez Gene ID: | 161931 |
Gene Name: | adenosine deaminase domain containing 2 |
Gene Aliases: |
TENRL |
Location: |
Chr.16:84191117-84197166 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADAD2 | NM_001145400.1 | NP_001138872.1 | ||
NM_139174.3 | NP_631913.3 | |||
XM_005255814.1 | XP_005255871.1 | |||
XM_011522919.1 | XP_011521221.1 | |||
AF447586.1 | AAM22869.1 | |||
AK093046.1 | BAC04032.1 | |||
AK131102.1 | BAC85152.1 | |||
AK315171.1 | ||||
BC033491.1 | AAH33491.1 | |||
DB036324.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3639413 | Chr.16:84172318 - 84253359 on Build GRCh38 | Loss | KCNG4 DNAAF1 TAF1C ADAD2 |
nsv833309 | Chr.16:84076867 - 84248591 on Build GRCh38 | Loss | HSDL1 MBTPS1 LOC729887 KCNG4 DNAAF1 TAF1C ADAD2 |
nsv1065790 | Chr.16:83830329 - 84243597 on Build GRCh38 | Loss | HSDL1 MBTPS1 LOC729887 SLC38A8 LOC105371371 MLYCD KCNG4 NECAB2 DNAAF1 OSGIN1 TAF1C ADAD2 |
esv2761930 | Chr.16:84007420 - 84312461 on Build GRCh38 | Gain | HSDL1 WFDC1 MBTPS1 LOC729887 SLC38A8 KCNG4 DNAAF1 LOC105376777 TAF1C ADAD2 |
nsv523437 | Chr.16:84176132 - 84196770 on Build GRCh38 | Loss | DNAAF1 TAF1C ADAD2 |
esv3639414 | Chr.16:84176251 - 84352273 on Build GRCh38 | Loss | WFDC1 KCNG4 DNAAF1 LOC105376777 TAF1C ADAD2 |
nsv457597 | Chr.16:84182086 - 84215151 on Build GRCh38 | Loss | TAF1C ADAD2 |
esv3639412 | Chr.16:84170074 - 84221406 on Build GRCh38 | Loss | KCNG4 DNAAF1 TAF1C ADAD2 |
nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain | GINS2 FOXC2-AS1 EMC8 IRF8 MTHFSD LOC105371374 CRISPLD2 TAF1C MIR1910 LINC00917 LOC102724344 KLHL36 ATP2C2 GSE1 KCNG4 FAM92B MIR6774 KIAA0513 USP10 C16orf74 LOC101928557 LOC101928614 FOXL1 LOC440390 LOC105376777 LOC101928474 LOC146513 COX4I1 LINC00311 LINC01082 MIR5093 WFDC1 TLDC1 FOXF1 LOC400548 LINC01081 FOXC2 DNAAF1 COTL1 FENDRR ADAD2 FLJ30679 ZDHHC7 |
nsv1059227 | Chr.16:84029323 - 84233403 on Build GRCh38 | Loss | HSDL1 MBTPS1 LOC729887 SLC38A8 KCNG4 DNAAF1 TAF1C ADAD2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |