Assay Details
Target Gene Details
Entrez Gene ID: | 9625 |
Gene Name: | apoptosis associated tyrosine kinase |
Gene Aliases: |
AATYK, AATYK1, LMR1, LMTK1, PPP1R77, p35BP |
Location: |
Chr.17:81117295-81166072 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK | NM_001080395.2 | NP_001073864.2 | ||
XM_006722193.2 | XP_006722256.1 | |||
XM_006722194.2 | XP_006722257.1 | |||
AK131529.1 | BAD18667.1 |
Target Gene Details
Entrez Gene ID: | 388428 |
Gene Name: | AATK antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:81165507-81183164 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK-AS1 | NR_027255.1 | 1 | 120 | |
AK126811.1 | 1 | 120 | BAC86704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | MIR6786 LOC100129503 CCDC137 LINC00482 CHMP6 BAIAP2-AS1 BAHCC1 SLC25A10 SLC38A10 FAAP100 LOC107985021 MIR3065 MIR338 ACTG1 BAIAP2 ARL16 CEP131 HGS LOC400627 LOC105371925 OXLD1 MIR1250 TMEM105 LOC101928855 RPTOR AATK-AS1 MIR657 FSCN2 LOC105371921 MRPL12 LOC100130370 PDE6G C17orf89 NPLOC4 AATK LOC107985022 MIR3186 TEPSIN MIR4740 TSPAN10 |
nsv1057306 | Chr.17:81154954 - 81207813 on Build GRCh38 | Loss | CEP131 AATK AATK-AS1 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | MIR6786 CCDC137 LINC00482 LRRC45 SLC25A10 C17orf62 SLC38A10 MAFG-AS1 FAAP100 UTS2R RAB40B ACTG1 SIRT7 FOXK2 FASN CEP131 LOC105371925 DCXR CD7 MCRIP1 TMEM105 CCDC57 SECTM1 ASPSCR1 P4HB OGFOD3 ANAPC11 AATK-AS1 MRPL12 TEX19 STRA13 NPLOC4 ALYREF LOC105376790 B3GNTL1 RAC3 TEPSIN MIR4740 TSPAN10 BAHCC1 NPB MIR4525 HEXDC PYCR1 CSNK1D GCGR ARL16 NARF MAFG HGS LOC105376791 OXLD1 PCYT2 NOTUM LOC101929552 DUS1L ARHGDIA SNORD134 LOC101929511 FSCN2 TBCD LOC105371944 LOC100130370 ZNF750 PDE6G GPS1 FN3KRP RFNG C17orf89 AATK MYADML2 PPP1R27 WDR45B MIR3186 FN3K SLC16A3 MIR6787 |
dgv46n68 | Chr.17:81133921 - 81288932 on Build GRCh38 | Loss | C17orf89 CEP131 AATK AATK-AS1 LOC105371925 SLC38A10 TEPSIN |
nsv1065052 | Chr.17:81080876 - 81204920 on Build GRCh38 | Gain | CEP131 AATK AATK-AS1 MIR657 MIR3065 MIR1250 MIR338 BAIAP2 |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | LINC00482 BAIAP2-AS1 AATK-AS1 MIR657 SLC38A10 MIR3065 MIR338 BAIAP2 C17orf89 CEP131 AATK LOC105371925 MIR1250 TEPSIN TMEM105 |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | BAIAP2-AS1 CEP131 AATK AATK-AS1 MIR657 MIR3065 MIR1250 MIR338 BAIAP2 |
nsv952389 | Chr.17:81137501 - 81593774 on Build GRCh38 | Deletion | LINC00482 BAHCC1 AATK-AS1 SLC38A10 FAAP100 FSCN2 LOC100130370 ACTG1 C17orf89 NPLOC4 CEP131 AATK LOC105371925 MIR3186 TEPSIN TMEM105 MIR4740 |
nsv1066947 | Chr.17:81094837 - 81181290 on Build GRCh38 | Loss | AATK AATK-AS1 MIR657 MIR3065 MIR1250 MIR338 BAIAP2 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |