Assay Details
Target Gene Details
Entrez Gene ID: | 283337 |
Gene Name: | zinc finger protein 740 |
Gene Aliases: |
Zfp740 |
Location: |
Chr.12:53180715-53190870 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF740 | NM_001004304.3 | 6 | 834 | NP_001004304.1 |
XM_006719344.2 | 6 | 809 | XP_006719407.1 | |
XM_011538194.2 | 6 | 1034 | XP_011536496.1 | |
XM_017019181.1 | 6 | 1068 | XP_016874670.1 | |
AK091946.1 | 3 | 217 | ||
AK292744.1 | 6 | 885 | ||
BC053557.1 | 6 | 837 | AAH53557.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832417 | Chr.12:53075874 - 53254231 on Build GRCh38 | Gain | IGFBP6 SPRYD3 ZNF740 MFSD5 SOAT2 CSAD ITGB7 RARG |
nsv558994 | Chr.12:53103618 - 53277765 on Build GRCh38 | Loss | ESPL1 ZNF740 MFSD5 SOAT2 CSAD ITGB7 RARG |
dgv2645n54 | Chr.12:53099603 - 53227927 on Build GRCh38 | Loss | IGFBP6 ZNF740 SOAT2 CSAD ITGB7 RARG |
nsv517066 | Chr.12:53006394 - 53340722 on Build GRCh38 | Loss | C12orf10 TNS2 LOC283335 AAAS ZNF740 EIF4B SOAT2 SP7 ITGB7 IGFBP6 SPRYD3 ESPL1 PFDN5 MFSD5 MIR6757 CSAD RARG |
dgv2644n54 | Chr.12:53064109 - 53211560 on Build GRCh38 | Loss | TNS2 IGFBP6 SPRYD3 ZNF740 SOAT2 CSAD ITGB7 RARG |
nsv469404 | Chr.12:53124036 - 53227927 on Build GRCh38 | Loss | ZNF740 SOAT2 CSAD ITGB7 RARG |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |