Assay Details
Target Gene Details
Entrez Gene ID: | 2271 |
Gene Name: | fumarate hydratase |
Gene Aliases: |
FMRD, HLRCC, LRCC, MCL, MCUL1 |
Location: |
Chr.1:241497557-241519785 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FH | NM_000143.3 | NP_000134.2 | ||
XM_011544132.2 | XP_011542434.1 | |||
AK312415.1 | ||||
BC003108.1 | AAH03108.1 | |||
BC017444.1 | AAH17444.1 | |||
BT009839.1 | AAP88841.1 | |||
DA129148.1 | ||||
KU178071.1 | ||||
KU178072.1 | ||||
KU178073.1 | ||||
KU178074.1 | ||||
KU178075.1 | ||||
KU178076.1 | ||||
M15502.1 | AAA52483.1 | |||
U48857.1 | AAD00071.1 | |||
U59309.1 | AAB66354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv549460 | Chr.1:241331278 - 241669985 on Build GRCh38 | Gain | FH CHML WDR64 RGS7 OPN3 KMO |
nsv1002214 | Chr.1:241295639 - 241936488 on Build GRCh38 | Gain | FH CHML WDR64 RGS7 OPN3 EXO1 KMO |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |