Assay Details
Target Gene Details
Entrez Gene ID: | 7917 |
Gene Name: | BCL2 associated athanogene 6 |
Gene Aliases: |
BAG-6, BAT3, D6S52E, G3 |
Location: |
Chr.6:31639028-31660900 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BAG6 | XM_017011279.1 | XP_016866768.1 |
Target Gene Details
Entrez Gene ID: | 57827 |
Gene Name: | chromosome 6 open reading frame 47 |
Gene Aliases: |
D6S53E, G4, NG34 |
Location: |
Chr.6:31658298-31660772 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C6orf47 | NM_021184.3 | 1 | 677 | NP_067007.3 |
AJ245433.1 | 1 | 638 | CAB51603.1 | |
AK315349.1 | 1 | 366 | ||
BC012950.1 | 1 | 677 | AAH12950.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv462871 | Chr.6:31607499 - 31664357 on Build GRCh38 | Loss | BAG6 C6orf47 PRRC2A APOM SNORA38 AIF1 MIR6832 GPANK1 |
nsv1126749 | Chr.6:31457323 - 32594423 on Build GRCh38 | Deletion | HSPA1L NFKBIL1 PBX2 CSNK2B MSH5-SAPCD1 ATP6V1G2-DDX39B FKBPL LTB HCG23 EHMT2 LY6G5C AGER LOC100287329 GPSM3 HSPA1B DDX39B STK19 TNXB LOC102725068 NCR3 PPT2 C2 LY6G6C LY6G5B C4B ATP6V1G2 MIR1236 LST1 CYP21A1P DDAH2 RNF5 TNF SNORD84 HLA-DRB1 LTA MIR6721 SAPCD1 SNORD52 C6orf47 PRRC2A VWA7 LOC101929163 AIF1 MIR6832 GPANK1 NOTCH4 DXO C6orf48 BAG6 MIR6833 VARS HLA-DRB6 DDX39B-AS1 SLC44A4 BTNL2 NELFE AGPAT1 PRRT1 EGFL8 CYP21A2 SNORD48 CFB HLA-DRB5 NEU1 SKIV2L MICB C4A LOC100507547 HCG26 C6orf25 MSH5 C2-AS1 LSM2 LY6G6D APOM LY6G6F TNXA LY6G6E SNORD117 MCCD1 PPT2-EGFL8 C6orf10 ABHD16A HSPA1A ZBTB12 ATF6B HLA-DRA SNORA38 CLIC1 SAPCD1-AS1 MIR4646 HCP5 |
nsv1073969 | Chr.6:31456422 - 33480523 on Build GRCh38 | Deletion | NFKBIL1 PSMB9 CSNK2B MSH5-SAPCD1 ATP6V1G2-DDX39B FKBPL LTB HCG23 BRD2 LY6G5C MIR6834 GPSM3 HSPA1B ZBTB9 MIR6873 TNXB LOC102725068 PPP1R2P1 PSMB8-AS1 LY6G6C LY6G5B C4B ATP6V1G2 MIR1236 HLA-DQB2 CYP21A1P DDAH2 B3GALT4 PSMB8 LOC100294145 SNORD84 SNORD52 C6orf47 RGL2 VWA7 AIF1 DXO BAG6 VARS DDX39B-AS1 BTNL2 AGPAT1 EGFL8 SNORD48 NEU1 SKIV2L C4A CUTA TAP1 C6orf25 MSH5 C2-AS1 LSM2 LY6G6D LY6G6E HLA-DPB2 SNORD117 MCCD1 SLC39A7 ABHD16A RING1 HLA-DOB MIR3135B HLA-DRA CLIC1 MIR4646 KIFC1 HCP5 HSPA1L MIR219A1 WDR46 PBX2 SYNGAP1 EHMT2 AGER LOC100287329 ZBTB22 DDX39B STK19 HLA-DPB1 NCR3 MIR5004 HLA-DMB PPT2 C2 LST1 TAPBP RNF5 TNF RXRB HLA-DRB1 LTA MIR6721 SAPCD1 PRRC2A LOC101929163 VPS52 MIR6832 GPANK1 NOTCH4 HLA-DQA2 C6orf48 MIR6833 HLA-DRB6 HCG25 HLA-DOA SLC44A4 NELFE PRRT1 TAP2 COL11A2 CYP21A2 PFDN6 HCG24 CFB PHF1 HLA-DRB5 HLA-DMA HLA-DQB1-AS1 MICB LOC100507547 RPS18 HCG26 APOM LY6G6F TNXA HLA-DQB1 DAXX PPT2-EGFL8 HSD17B8 C6orf10 HSPA1A ZBTB12 ATF6B SNORA38 HLA-DPA1 SAPCD1-AS1 HLA-DQA1 |
dgv10403n54 | Chr.6:31312324 - 32049763 on Build GRCh38 | Loss | HSPA1L NFKBIL1 CSNK2B MSH5-SAPCD1 ATP6V1G2-DDX39B LTB EHMT2 LY6G5C LOC100287329 HSPA1B DDX39B STK19 MIR6891 TNXB LOC102725068 NCR3 MICA C2 LY6G6C LY6G5B C4B ATP6V1G2 MIR1236 LST1 CYP21A1P DDAH2 TNF SNORD84 LTA SAPCD1 SNORD52 C6orf47 PRRC2A HLA-B VWA7 AIF1 MIR6832 GPANK1 DXO C6orf48 BAG6 VARS DDX39B-AS1 SLC44A4 NELFE CYP21A2 SNORD48 CFB NEU1 SKIV2L MICB C4A HCG26 C6orf25 MSH5 C2-AS1 LSM2 LY6G6D APOM LY6G6F TNXA LY6G6E SNORD117 MCCD1 ABHD16A HSPA1A ZBTB12 SNORA38 CLIC1 SAPCD1-AS1 MIR4646 HCP5 |
nsv1112900 | Chr.6:31455323 - 33480423 on Build GRCh38 | Deletion | NFKBIL1 PSMB9 CSNK2B MSH5-SAPCD1 ATP6V1G2-DDX39B FKBPL LTB HCG23 BRD2 LY6G5C MIR6834 GPSM3 HSPA1B ZBTB9 MIR6873 TNXB LOC102725068 PPP1R2P1 PSMB8-AS1 LY6G6C LY6G5B C4B ATP6V1G2 MIR1236 HLA-DQB2 CYP21A1P DDAH2 B3GALT4 PSMB8 LOC100294145 SNORD84 SNORD52 C6orf47 RGL2 VWA7 AIF1 DXO BAG6 VARS DDX39B-AS1 BTNL2 AGPAT1 EGFL8 SNORD48 NEU1 SKIV2L C4A CUTA TAP1 C6orf25 MSH5 C2-AS1 LSM2 LY6G6D LY6G6E HLA-DPB2 SNORD117 MCCD1 SLC39A7 ABHD16A RING1 HLA-DOB MIR3135B HLA-DRA CLIC1 MIR4646 KIFC1 HCP5 HSPA1L MIR219A1 WDR46 PBX2 SYNGAP1 EHMT2 AGER LOC100287329 ZBTB22 DDX39B STK19 HLA-DPB1 NCR3 MIR5004 HLA-DMB PPT2 C2 LST1 TAPBP RNF5 TNF RXRB HLA-DRB1 LTA MIR6721 SAPCD1 PRRC2A LOC101929163 VPS52 MIR6832 GPANK1 NOTCH4 HLA-DQA2 C6orf48 MIR6833 HLA-DRB6 HCG25 HLA-DOA SLC44A4 NELFE PRRT1 TAP2 COL11A2 CYP21A2 PFDN6 HCG24 CFB PHF1 HLA-DRB5 HLA-DMA HLA-DQB1-AS1 MICB LOC100507547 RPS18 HCG26 APOM LY6G6F TNXA HLA-DQB1 DAXX PPT2-EGFL8 HSD17B8 C6orf10 HSPA1A ZBTB12 ATF6B SNORA38 HLA-DPA1 SAPCD1-AS1 HLA-DQA1 |
dgv10404n54 | Chr.6:31312550 - 32516591 on Build GRCh38 | Loss | HSPA1L NFKBIL1 PBX2 CSNK2B MSH5-SAPCD1 ATP6V1G2-DDX39B FKBPL LTB HCG23 EHMT2 LY6G5C AGER LOC100287329 GPSM3 HSPA1B DDX39B STK19 MIR6891 TNXB LOC102725068 NCR3 MICA PPT2 C2 LY6G6C LY6G5B C4B ATP6V1G2 MIR1236 LST1 CYP21A1P DDAH2 RNF5 TNF SNORD84 LTA MIR6721 SAPCD1 SNORD52 C6orf47 PRRC2A HLA-B VWA7 LOC101929163 AIF1 MIR6832 GPANK1 NOTCH4 DXO C6orf48 BAG6 MIR6833 VARS DDX39B-AS1 SLC44A4 BTNL2 NELFE AGPAT1 PRRT1 EGFL8 CYP21A2 SNORD48 CFB NEU1 SKIV2L MICB C4A LOC100507547 HCG26 C6orf25 MSH5 C2-AS1 LSM2 LY6G6D APOM LY6G6F TNXA LY6G6E SNORD117 MCCD1 PPT2-EGFL8 C6orf10 ABHD16A HSPA1A ZBTB12 ATF6B HLA-DRA SNORA38 CLIC1 SAPCD1-AS1 MIR4646 HCP5 |
nsv1161370 | Chr.6:31600692 - 31714252 on Build GRCh38 | Deletion | LY6G5B BAG6 APOM LY6G6F CSNK2B LY6G6E LY6G5C C6orf47 PRRC2A ABHD16A SNORA38 AIF1 MIR6832 MIR4646 GPANK1 |
nsv830627 | Chr.6:31528437 - 31692173 on Build GRCh38 | Loss | BAG6 NFKBIL1 CSNK2B DDX39B-AS1 ATP6V1G2-DDX39B LTB LY6G5C LOC100287329 DDX39B NCR3 LY6G5B ATP6V1G2 LST1 APOM SNORD117 TNF SNORD84 MCCD1 LTA C6orf47 PRRC2A ABHD16A SNORA38 AIF1 MIR6832 GPANK1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |