Assay Details
Target Gene Details
Entrez Gene ID: | 574407 |
Gene Name: | chromosome 1 open reading frame 145 |
Gene Aliases: |
- |
Location: |
Chr.1:228203506-228213664 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C1orf145 | NR_073154.1 | |||
NR_073155.1 | ||||
AK056556.1 | BAB71215.1 |
Target Gene Details
Entrez Gene ID: | 84033 |
Gene Name: | obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF |
Gene Aliases: |
ARHGEF30, UNC89 |
Location: |
Chr.1:228208013-228381431 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1069355 | Chr.1:228212898 - 228214099 on Build GRCh38 | Deletion | C1orf145 OBSCN |
nsv1143478 | Chr.1:228212799 - 228214299 on Build GRCh38 | Deletion | C1orf145 OBSCN |
nsv1160102 | Chr.1:228081212 - 228239280 on Build GRCh38 | Deletion | IBA57-AS1 C1orf145 MIR3620 IBA57 OBSCN MRPL55 GUK1 GJC2 C1orf35 ARF1 |
nsv826908 | Chr.1:227707685 - 228503399 on Build GRCh38 | Gain | C1orf145 LOC105373289 GJC2 MIR4666A LOC101927401 IBA57-AS1 HIST3H2BB WNT3A LOC101060022 LOC100506571 IBA57 RNF187 MRPL55 SNAP47 PRSS38 GUK1 JMJD4 MIR5008 HIST3H2A LOC107985355 TRIM17 WNT9A ARF1 MIR6742 TRIM11 MIR3620 OBSCN HIST3H3 C1orf35 |
esv32853 | Chr.1:227692415 - 228670462 on Build GRCh38 | Gain | RNA5S11 C1orf145 LOC105373289 RNA5S3 DUSP5P1 LOC101927401 HIST3H2BB LOC101060022 LOC100506571 IBA57 BTNL10 MRPL55 SNAP47 PRSS38 GUK1 RNA5S6 JMJD4 RNA5S2 RNA5S5 HIST3H2A RNA5S4 TRIM17 WNT9A ARF1 MIR6742 RNA5S7 RNA5S14 RHOU RNA5S17 MIR3620 OBSCN LOC105373132 HIST3H3 C1orf35 RNA5S10 GJC2 MIR4666A RNA5S15 RNA5S13 RNA5S12 IBA57-AS1 WNT3A RNA5S1 RNF187 MIR5008 RNA5S16 LOC107985355 RNA5S9 TRIM11 RNA5S8 |
nsv827019 | Chr.1:228126466 - 228463319 on Build GRCh38 | Loss | C1orf145 HIST3H2A GJC2 MIR4666A LOC101927401 TRIM17 MIR6742 TRIM11 IBA57-AS1 HIST3H2BB IBA57 OBSCN GUK1 HIST3H3 |
nsv952138 | Chr.1:228207400 - 228217099 on Build GRCh38 | Deletion | C1orf145 OBSCN |
nsv523935 | Chr.1:227850423 - 228494048 on Build GRCh38 | Loss | C1orf145 GJC2 MIR4666A LOC101927401 IBA57-AS1 HIST3H2BB WNT3A LOC101060022 LOC100506571 IBA57 RNF187 MRPL55 GUK1 MIR5008 HIST3H2A LOC107985355 TRIM17 WNT9A ARF1 MIR6742 TRIM11 MIR3620 OBSCN HIST3H3 C1orf35 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |