Assay Details
Target Gene Details
Entrez Gene ID: | 134637 |
Gene Name: | adenosine deaminase, tRNA specific 2 |
Gene Aliases: |
DEADC1, TAD2, dJ20N2, dJ20N2.1 |
Location: |
Chr.6:143422832-143450750 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADAT2 | NM_001286259.1 | 6 | 2124 | NP_001273188.1 |
NM_182503.2 | 6 | 1026 | NP_872309.2 | |
XM_011535439.2 | 9 | 1182 | XP_011533741.1 | |
XM_011535440.1 | 9 | 3914 | XP_011533742.1 | |
XM_011535444.2 | 8 | 1045 | XP_011533746.1 | |
XM_017010260.1 | 7 | 4443 | XP_016865749.1 | |
XM_017010261.1 | 6 | 4400 | XP_016865750.1 | |
XM_017010262.1 | 6 | 904 | XP_016865751.1 | |
XM_017010263.1 | 5 | 878 | XP_016865752.1 | |
AK126201.1 | 6 | 2130 | ||
BC037955.1 | 6 | 986 | ||
BX538182.1 | 6 | 1424 | CAD98054.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |