Assay Details
Target Gene Details
Entrez Gene ID: | 65220 |
Gene Name: | NAD kinase |
Gene Aliases: |
dJ283E3.1 |
Location: |
Chr.1:1751232-1780514 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1003545 | Chr.1:1696467 - 1760882 on Build GRCh38 | Gain+Loss | MMP23A NADK SLC35E2 CDK11A |
dgv5n111 | Chr.1:1575421 - 1823403 on Build GRCh38 | Deletion | MMP23A GNB1 NADK MMP23B CDK11B SLC35E2 CDK11A FNDC10 SLC35E2B MIB2 |
nsv834091 | Chr.1:1701359 - 1880638 on Build GRCh38 | Loss | MMP23A GNB1 NADK SLC35E2 CDK11A |
nsv545041 | Chr.1:1753033 - 1780259 on Build GRCh38 | Loss | NADK |
nsv1160797 | Chr.1:1566216 - 1805918 on Build GRCh38 | Duplication | SSU72 MMP23A GNB1 NADK MMP23B CDK11B SLC35E2 CDK11A FNDC10 SLC35E2B MIB2 |
nsv482937 | Chr.1:10001 - 2368561 on Build GRCh38 | Loss | SSU72 UBE2J2 MIR1302-2 FAM87B AURKAIP1 TMEM88B DDX11L1 MRPL20 ACAP3 CFAP74 LOC100288069 LOC100133331 AGRN FNDC10 TMEM52 CPSF3L LOC107984841 PUSL1 MMP23B MIR6723 TTLL10 LOC102725121 C1orf159 CDK11A MIR6859-2 FAM41C LOC107985728 ATAD3A OR4F29 DVL1 FAM132A MIR6808 CDK11B LOC105378592 MTND2P28 LOC105378947 LOC105378948 LOC100129534 ATAD3C MIR429 MTND1P23 LOC101928626 VWA1 CCNL2 LOC100506504 MIR6859-1 MORN1 LOC100288175 SLC35E2B PLEKHN1 ATAD3B LOC100134822 SKI MIR6726 LOC148413 GABRD LOC100132287 MIB2 LOC107985729 LOC100130417 TMEM240 MXRA8 NOC2L GNB1 SCNN1D LOC102724312 SAMD11 MIR200A CPTP CALML6 LOC105378949 B3GALT6 WASH7P OR4F16 LOC105378591 KLHL17 FAM138A LOC105378589 TAS1R3 MIR200B SLC35E2 PRKCZ LOC729737 TTLL10-AS1 HES4 MMP23A PERM1 TNFRSF4 LOC100287934 LINC01128 LINC00115 LINC01342 RNF223 SDF4 LOC284600 FAAP20 ISG15 NADK MIR6727 ANKRD65 TNFRSF18 OR4F5 |
esv2760410 | Chr.1:1565561 - 1756248 on Build GRCh38 | Gain+Loss | SSU72 MMP23A NADK MMP23B CDK11B SLC35E2 CDK11A FNDC10 SLC35E2B MIB2 |
nsv950521 | Chr.1:1729062 - 1760461 on Build GRCh38 | Deletion | NADK SLC35E2 |
nsv945749 | Chr.1:1751857 - 1760427 on Build GRCh38 | Duplication | NADK |
nsv545030 | Chr.1:1689422 - 1774697 on Build GRCh38 | Gain | MMP23A NADK SLC35E2 CDK11A SLC35E2B |
nsv10161 | Chr.1:776731 - 1777210 on Build GRCh38 | Gain+Loss | LOC100130417 SSU72 UBE2J2 FAM87B TMEM240 AURKAIP1 MXRA8 NOC2L TMEM88B MRPL20 SCNN1D LOC102724312 SAMD11 ACAP3 LOC100288069 MIR200A AGRN FNDC10 CPSF3L CPTP PUSL1 MMP23B B3GALT6 TTLL10 C1orf159 CDK11A KLHL17 FAM41C LOC107985728 ATAD3A DVL1 FAM132A MIR6808 TAS1R3 MIR200B CDK11B SLC35E2 LOC105378948 TTLL10-AS1 ATAD3C HES4 MIR429 MMP23A PERM1 VWA1 CCNL2 TNFRSF4 LOC100287934 LINC01128 LOC100288175 SLC35E2B PLEKHN1 LINC00115 LINC01342 ATAD3B RNF223 SDF4 LOC284600 ISG15 NADK MIR6727 MIR6726 LOC148413 ANKRD65 TNFRSF18 MIB2 LOC107985729 |
dgv4e212 | Chr.1:1696966 - 1756488 on Build GRCh38 | Gain | MMP23A NADK SLC35E2 CDK11A |
esv2758913 | Chr.1:1389149 - 2092899 on Build GRCh38 | Gain+Loss | SSU72 LOC105378589 CDK11B SLC35E2 TMEM240 TMEM88B GNB1 PRKCZ MRPL20 LOC102724312 CFAP74 FNDC10 ATAD3C TMEM52 MMP23A CALML6 VWA1 CCNL2 MMP23B LOC105378949 CDK11A SLC35E2B ATAD3B LOC105378591 NADK LOC148413 ANKRD65 ATAD3A GABRD MIB2 LOC107985729 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |