Assay Details
Target Gene Details
Entrez Gene ID: | 54869 |
Gene Name: | EPS8 like 1 |
Gene Aliases: |
DRC3, EPS8R1, PP10566 |
Location: |
Chr.19:55075853-55087923 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPS8L1 | NM_017729.3 | 7 | 873 | NP_060199.3 |
NM_133180.2 | 12 | 1256 | NP_573441.2 | |
XM_005259020.1 | 12 | 1256 | XP_005259077.1 | |
XM_011527050.1 | 7 | 873 | XP_011525352.1 | |
XM_011527051.2 | 12 | 1256 | XP_011525353.1 | |
XM_011527052.2 | 12 | 1256 | XP_011525354.1 | |
AF282167.1 | 3 | 405 | AAG03038.1 | |
AF370395.1 | 12 | 1208 | AAQ15231.1 | |
AK000265.1 | 7 | 873 | BAA91041.1 | |
AK057052.1 | 3 | 553 | ||
AK075098.1 | 12 | 1207 | BAC11399.1 | |
AK296732.1 | 10 | 1106 | ||
AK301615.1 | 2 | 216 | ||
AK309305.1 | 12 | 1256 | ||
AK309908.1 | 10 | 1182 | ||
AY074928.1 | 11 | 1209 | AAL76117.1 | |
BC004907.2 | 9 | 1276 | AAH04907.2 | |
BC015763.1 | 7 | 842 | AAH15763.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv817874 | Chr.19:55067334 - 55103555 on Build GRCh38 | Loss | PPP1R12C RDH13 EPS8L1 |
esv3644829 | Chr.19:55030893 - 55138732 on Build GRCh38 | Gain | PPP1R12C MIR7975 GP6 RDH13 EPS8L1 TNNT1 |
esv2667609 | Chr.19:54472514 - 55094226 on Build GRCh38 | Deletion | KIR3DX1 FCAR EPS8L1 KIR2DL3 LILRA2 NLRP2 PPP1R12C CDC42EP5 KIR3DL3 LILRB1 LOC102724300 KIR3DL2 KIR2DL1 NCR1 KIR3DL1 MIR8061 LAIR2 LILRB4 KIR2DS4 NLRP7 LOC101928804 GP6 RDH13 LILRP2 LILRA1 KIR2DL4 |
esv3893251 | Chr.19:55079303 - 55094733 on Build GRCh38 | Loss | PPP1R12C EPS8L1 |
esv2758772 | Chr.19:55026094 - 55139319 on Build GRCh38 | Gain | PPP1R12C MIR7975 GP6 RDH13 EPS8L1 TNNT1 |
nsv817875 | Chr.19:55070143 - 55090747 on Build GRCh38 | Loss | EPS8L1 |
esv2718872 | Chr.19:54472594 - 55094247 on Build GRCh38 | Deletion | KIR3DX1 FCAR EPS8L1 KIR2DL3 LILRA2 NLRP2 PPP1R12C CDC42EP5 KIR3DL3 LILRB1 LOC102724300 KIR3DL2 KIR2DL1 NCR1 KIR3DL1 MIR8061 LAIR2 LILRB4 KIR2DS4 NLRP7 LOC101928804 GP6 RDH13 LILRP2 LILRA1 KIR2DL4 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |