Assay Details
Target Gene Details
Entrez Gene ID: | 283849 |
Gene Name: | exocyst complex component 3 like 1 |
Gene Aliases: |
EXOC3L |
Location: |
Chr.16:67184379-67190204 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXOC3L1 | NM_178516.3 | 5 | 732 | NP_848611.2 |
AK092858.1 | 5 | 624 | BAC03991.1 | |
AK092906.1 | 5 | 496 | BAC04000.1 | |
AK292004.1 | 5 | 641 | ||
AK300915.1 | 5 | 517 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | FAM65A LRRC29 HSD11B2 EXOC3L1 ELMO3 KCTD19 PSKH1 PARD6A ACD ATP6V0D1 CTRL FBXL8 THAP11 CARMIL2 C16orf86 TRADD NUTF2 PLEKHG4 TSNAXIP1 AGRP SLC9A5 RANBP10 TMEM208 EDC4 LOC100505942 NRN1L DDX28 DPEP2 DUS2 HSF4 MIR328 LOC107984813 FHOD1 LCAT DPEP3 LOC105369155 CTCF CENPT GFOD2 ZDHHC1 E2F4 KIAA0895L LRRC36 TPPP3 LOC100131303 ENKD1 PSMB10 SLC12A4 NOL3 |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss | LOC105369155 LRRC29 TRADD PLEKHG4 HSF4 EXOC3L1 ELMO3 SLC9A5 MIR328 E2F4 KIAA0895L TMEM208 FHOD1 FBXL8 B3GNT9 C16orf70 NOL3 |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion | LOC105369155 LRRC29 TRADD HSF4 EXOC3L1 ELMO3 MIR328 E2F4 KIAA0895L TMEM208 FHOD1 FBXL8 B3GNT9 C16orf70 NOL3 |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion | KIAA0895L LOC105369155 LRRC29 HSF4 EXOC3L1 FBXL8 ELMO3 NOL3 MIR328 E2F4 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | FAM65A LRRC29 HSD11B2 EXOC3L1 ELMO3 KCTD19 PSKH1 PARD6A ACD ATP6V0D1 CTRL FBXL8 THAP11 CARMIL2 C16orf86 TRADD NUTF2 PLEKHG4 TSNAXIP1 AGRP SLC9A5 RANBP10 TMEM208 EDC4 LOC100505942 C16orf70 NRN1L DDX28 DPEP2 DUS2 HSF4 MIR328 LOC107984813 FHOD1 LCAT B3GNT9 DPEP3 LOC105369155 CTCF CENPT GFOD2 ZDHHC1 E2F4 KIAA0895L LRRC36 TPPP3 LOC100131303 ENKD1 PSMB10 SLC12A4 NOL3 |
nsv457513 | Chr.16:67181417 - 67282697 on Build GRCh38 | Loss | KIAA0895L LOC105369155 TMEM208 LRRC29 FHOD1 PLEKHG4 EXOC3L1 ELMO3 SLC9A5 MIR328 E2F4 |
nsv524492 | Chr.16:67174030 - 67294550 on Build GRCh38 | Loss | LOC105369155 LRRC29 PLEKHG4 EXOC3L1 ELMO3 SLC9A5 KCTD19 MIR328 E2F4 KIAA0895L TMEM208 FHOD1 NOL3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |