Assay Details
Target Gene Details
Entrez Gene ID: | 94101 |
Gene Name: | ORMDL sphingolipid biosynthesis regulator 1 |
Gene Aliases: |
- |
Location: |
Chr.2:189766149-189784371 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ORMDL1 | NM_001128150.1 | 4 | 795 | NP_001121622.1 |
NM_016467.4 | 5 | 905 | NP_057551.1 | |
XM_005246970.2 | 4 | 824 | XP_005247027.1 | |
XM_011512199.2 | XP_011510501.1 | |||
XM_017005347.1 | 5 | 944 | XP_016860836.1 | |
XM_017005348.1 | XP_016860837.1 | |||
AB064959.1 | 3 | 1364 | BAC11710.1 | |
AB064960.1 | 4 | 803 | BAC11711.1 | |
AF151036.1 | 4 | 743 | AAF36122.1 | |
AF395704.1 | 5 | 860 | AAM43503.1 | |
AK074756.1 | 4 | 794 | BAC11184.1 | |
AK075160.1 | 3 | 796 | BAC11441.1 | |
AK126336.1 | 1 | 3817 | ||
BC005200.1 | 4 | 762 | AAH05200.1 | |
CR936635.1 | 3 | 1472 | CAI56776.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |