Assay Details
Target Gene Details
Entrez Gene ID: | 79885 |
Gene Name: | histone deacetylase 11 |
Gene Aliases: |
HD11 |
Location: |
Chr.3:13480171-13506424 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HDAC11 | NM_001136041.2 | 2 | 175 | NP_001129513.1 |
NM_024827.3 | 2 | 207 | NP_079103.2 | |
XM_005265491.1 | 2 | 172 | XP_005265548.1 | |
XM_005265492.1 | 2 | 251 | XP_005265549.1 | |
XM_006713338.1 | 2 | 207 | XP_006713401.1 | |
XM_011534131.1 | 2 | 295 | XP_011532433.1 | |
XM_011534132.2 | 2 | 97 | XP_011532434.2 | |
XM_011534135.1 | 2 | 115 | XP_011532437.1 | |
XM_017007234.1 | 2 | 295 | XP_016862723.1 | |
AK025426.1 | 2 | 98 | BAB15127.1 | |
AK025890.1 | 2 | 98 | BAB15272.1 | |
AK293223.1 | 2 | 108 | ||
AK299021.1 | 2 | 116 | ||
AL834223.1 | 2 | 465 | CAH56375.1 | |
BC009676.1 | 1 | 83 | AAH09676.1 | |
BG708402.1 | 2 | 177 | ||
DB025431.1 | 2 | 207 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |