Assay Details
Target Gene Details
Entrez Gene ID: | 2638 |
Gene Name: | GC, vitamin D binding protein |
Gene Aliases: |
DBP, DBP/GC, GRD3, Gc-MAF, GcMAF, HEL-S-51, VDBG, VDBP |
Location: |
Chr.4:71741693-71805520 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GC | NM_000583.3 | 7 | 1107 | NP_000574.2 |
NM_001204306.1 | 8 | 978 | NP_001191235.1 | |
NM_001204307.1 | 8 | 915 | NP_001191236.1 | |
XM_006714177.2 | 7 | 970 | XP_006714240.1 | |
AK223458.1 | 8 | 978 | BAD97178.1 | |
AK290827.1 | 7 | 817 | ||
AK298433.1 | 6 | 675 | ||
AK309595.1 | 8 | 915 | ||
AK312971.1 | 7 | 821 | ||
AK315853.1 | 7 | 970 | ||
BC057228.1 | 7 | 825 | AAH57228.1 | |
EU794674.1 | 7 | 916 | ||
X03178.1 | 7 | 916 | CAA26938.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829967 | Chr.4:71602549 - 71760380 on Build GRCh38 | Loss | GC |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |