Assay Details
Target Gene Details
Entrez Gene ID: | 662 |
Gene Name: | BCL2 interacting protein 1 |
Gene Aliases: |
NIP1, SEC20, TRG-8 |
Location: |
Chr.5:173144442-173164387 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BNIP1 | NM_001205.2 | NP_001196.2 | ||
NM_013978.2 | NP_053581.2 | |||
NM_013979.2 | NP_053582.2 | |||
NM_013980.2 | NP_053583.2 | |||
XM_011534638.1 | XP_011532940.1 | |||
XM_011534639.1 | XP_011532941.1 | |||
AF083956.1 | AAC33124.1 | |||
AF083957.1 | AAC33125.1 | |||
AF083958.1 | AAC33126.1 | |||
AU311051.1 | ||||
AY216799.1 | AAO91805.1 | |||
BC010959.1 | AAH10959.1 | |||
U15172.1 | AAC00020.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |