Assay Details
Target Gene Details
Entrez Gene ID: | 81889 |
Gene Name: | fumarylacetoacetate hydrolase domain containing 1 |
Gene Aliases: |
C16orf36, YISKL |
Location: |
Chr.16:1827224-1840207 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAHD1 | NM_001018104.2 | 3 | 1030 | NP_001018114.1 |
NM_001142398.1 | 2 | 911 | NP_001135870.1 | |
BC063017.1 | 2 | 915 | AAH63017.1 |
Target Gene Details
Entrez Gene ID: | 254528 |
Gene Name: | meiosis specific with OB domains |
Gene Aliases: |
C16orf73, gs129 |
Location: |
Chr.16:1833983-1872178 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MEIOB | NM_001163560.2 | NP_001157032.1 | ||
NM_152764.2 | NP_689977.2 | |||
BC029829.1 | AAH29829.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517196 | Chr.16:1729694 - 1844960 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv833119 | Chr.16:1834308 - 1911484 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv571175 | Chr.16:1806439 - 1875278 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv1040440 | Chr.16:1811739 - 1866102 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1051692 | Chr.16:1505070 - 1866102 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1042986 | Chr.16:1784723 - 1866102 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv457337 | Chr.16:1813519 - 1855005 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv571177 | Chr.16:1817039 - 1897015 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)