Assay Details
Target Gene Details
Entrez Gene ID: | 57591 |
Gene Name: | megakaryoblastic leukemia (translocation) 1 |
Gene Aliases: |
BSAC, MAL, MRTF-A |
Location: |
Chr.22:40410281-40636719 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MKL1 | NM_001282660.1 | NP_001269589.1 | ||
NM_001282661.1 | NP_001269590.1 | |||
NM_001282662.1 | NP_001269591.1 | |||
NM_001318139.1 | NP_001305068.1 | |||
NM_020831.4 | NP_065882.1 | |||
XM_005261692.1 | XP_005261749.1 | |||
XM_005261694.1 | XP_005261751.1 | |||
XM_011530283.1 | XP_011528585.1 | |||
XM_011530285.1 | XP_011528587.1 | |||
XM_011530287.1 | XP_011528589.1 | |||
XM_017028888.1 | XP_016884377.1 | |||
XM_017028889.1 | XP_016884378.1 | |||
AB037859.2 | ||||
AF368061.1 | AAK54721.1 | |||
AJ297257.1 | CAC38826.1 | |||
AJ297258.1 | CAC38827.1 | |||
AL713710.1 | CAD28507.2 | |||
BC115039.1 | ||||
HG764171.1 |
Target Gene Details
Entrez Gene ID: | 27352 |
Gene Name: | small G protein signaling modulator 3 |
Gene Aliases: |
MAP, RABGAP5, RUSC3, RUTBC3, RabGAP-5, rabGAPLP |
Location: |
Chr.22:40370530-40410289 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SGSM3 | NM_001301849.1 | NP_001288778.1 | ||
NM_015705.5 | NP_056520.2 | |||
XM_005261573.3 | XP_005261630.1 | |||
XM_005261574.4 | XP_005261631.1 | |||
XM_017028777.1 | XP_016884266.1 | |||
AW512418.1 | ||||
HY229847.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829233 | Chr.22:40404227 - 40438253 on Build GRCh38 | Loss | MKL1 SGSM3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |