Assay Details
Target Gene Details
Entrez Gene ID: | 6356 |
Gene Name: | C-C motif chemokine ligand 11 |
Gene Aliases: |
SCYA11 |
Location: |
Chr.17:34285668-34288180 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL11 | NM_002986.2 | 1 | 36 | NP_002977.1 |
BC017850.1 | AAH17850.1 | |||
BF197516.1 | 3 | 421 | ||
U46573.1 | AAA98957.1 | |||
Z69291.1 | CAA93258.1 | |||
Z75668.1 | CAA99997.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3141n100 | Chr.17:33630132 - 34611003 on Build GRCh38 | Gain | CCL1 LOC107987247 CCL8 CCL11 C17orf102 CCL7 TMEM132E CCL13 CCL2 LOC101927239 ASIC2 |
nsv517423 | Chr.17:33777656 - 34498242 on Build GRCh38 | Gain+Loss | CCL1 LOC107987247 CCL8 CCL11 CCL7 CCL13 CCL2 LOC101927239 ASIC2 |
nsv574714 | Chr.17:33640441 - 34608994 on Build GRCh38 | Gain | CCL1 LOC107987247 CCL8 CCL11 C17orf102 CCL7 TMEM132E CCL13 CCL2 LOC101927239 ASIC2 |
esv3572553 | Chr.17:33632208 - 34617133 on Build GRCh38 | Gain | CCL1 LOC107987247 CCL8 CCL11 C17orf102 CCL7 TMEM132E CCL13 CCL2 LOC101927239 ASIC2 |
nsv519297 | Chr.17:33640660 - 34608994 on Build GRCh38 | Gain | CCL1 LOC107987247 CCL8 CCL11 C17orf102 CCL7 TMEM132E CCL13 CCL2 LOC101927239 ASIC2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |