Assay Details
Target Gene Details
Entrez Gene ID: | 161931 |
Gene Name: | adenosine deaminase domain containing 2 |
Gene Aliases: |
TENRL |
Location: |
Chr.16:84191117-84197166 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADAD2 | NM_001145400.1 | NP_001138872.1 | ||
NM_139174.3 | NP_631913.3 | |||
XM_005255814.1 | XP_005255871.1 | |||
XM_011522919.1 | XP_011521221.1 | |||
XM_011522920.1 | XP_011521222.1 | |||
AF447586.1 | AAM22869.1 | |||
AK093046.1 | BAC04032.1 | |||
AK093523.1 | 1 | 1103 | ||
AK131102.1 | BAC85152.1 | |||
AK315171.1 | ||||
BC033491.1 | AAH33491.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3639413 | Chr.16:84172318 - 84253359 on Build GRCh38 | Loss | DNAAF1 TAF1C ADAD2 KCNG4 |
nsv518342 | Chr.16:84194593 - 84196629 on Build GRCh38 | Loss | ADAD2 |
nsv833309 | Chr.16:84076867 - 84248591 on Build GRCh38 | Loss | DNAAF1 TAF1C MBTPS1 ADAD2 KCNG4 LOC729887 HSDL1 |
nsv1065790 | Chr.16:83830329 - 84243597 on Build GRCh38 | Loss | DNAAF1 LOC105371371 SLC38A8 TAF1C MLYCD MBTPS1 ADAD2 KCNG4 LOC729887 NECAB2 HSDL1 OSGIN1 |
esv2761930 | Chr.16:84007420 - 84312461 on Build GRCh38 | Gain | DNAAF1 SLC38A8 TAF1C WFDC1 MBTPS1 ADAD2 KCNG4 LOC729887 LOC105376777 HSDL1 |
nsv523437 | Chr.16:84176132 - 84196770 on Build GRCh38 | Loss | DNAAF1 TAF1C ADAD2 |
esv3639414 | Chr.16:84176251 - 84352273 on Build GRCh38 | Loss | DNAAF1 TAF1C WFDC1 ADAD2 KCNG4 LOC105376777 |
nsv457597 | Chr.16:84182086 - 84215151 on Build GRCh38 | Loss | TAF1C ADAD2 |
esv3639412 | Chr.16:84170074 - 84221406 on Build GRCh38 | Loss | DNAAF1 TAF1C ADAD2 KCNG4 |
nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain | LOC101928474 LINC01081 TAF1C COX4I1 EMC8 LOC146513 LINC01082 C16orf74 LOC400548 FOXC2 KCNG4 GINS2 LOC101928614 FOXF1 COTL1 ATP2C2 MIR1910 MIR6774 LOC105371374 FLJ30679 TLDC1 DNAAF1 LOC440390 FAM92B FOXL1 LOC102724344 WFDC1 LOC101928557 KLHL36 IRF8 MTHFSD GSE1 ADAD2 CRISPLD2 MIR5093 KIAA0513 USP10 ZDHHC7 FOXC2-AS1 LINC00917 LINC00311 FENDRR LOC105376777 |
nsv1059227 | Chr.16:84029323 - 84233403 on Build GRCh38 | Loss | DNAAF1 SLC38A8 TAF1C MBTPS1 ADAD2 KCNG4 LOC729887 HSDL1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |