Assay Details
Target Gene Details
Entrez Gene ID: | 6355 |
Gene Name: | C-C motif chemokine ligand 8 |
Gene Aliases: |
HC14, MCP-2, MCP2, SCYA10, SCYA8 |
Location: |
Chr.17:34319047-34321402 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL8 | NM_005623.2 | 1 | 272 | NP_005614.2 |
BC126242.1 | 1 | 79 | ||
Y16645.1 | 1 | 289 | CAA76341.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3141n100 | Chr.17:33630132 - 34611003 on Build GRCh38 | Gain | CCL8 CCL1 CCL7 ASIC2 C17orf102 CCL2 LOC107987247 LOC101927239 CCL11 TMEM132E CCL13 |
nsv517423 | Chr.17:33777656 - 34498242 on Build GRCh38 | Gain+Loss | CCL8 CCL1 CCL7 ASIC2 CCL2 LOC107987247 LOC101927239 CCL11 CCL13 |
nsv574714 | Chr.17:33640441 - 34608994 on Build GRCh38 | Gain | CCL8 CCL1 CCL7 ASIC2 C17orf102 CCL2 LOC107987247 LOC101927239 CCL11 TMEM132E CCL13 |
esv3572553 | Chr.17:33632208 - 34617133 on Build GRCh38 | Gain | CCL8 CCL1 CCL7 ASIC2 C17orf102 CCL2 LOC107987247 LOC101927239 CCL11 TMEM132E CCL13 |
nsv519297 | Chr.17:33640660 - 34608994 on Build GRCh38 | Gain | CCL8 CCL1 CCL7 ASIC2 C17orf102 CCL2 LOC107987247 LOC101927239 CCL11 TMEM132E CCL13 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |