Assay Details
Target Gene Details
Entrez Gene ID: | 8091 |
Gene Name: | high mobility group AT-hook 2 |
Gene Aliases: |
BABL, HMGI-C, HMGIC, LIPO, STQTL9 |
Location: |
Chr.12:65824460-65966291 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HMGA2 | NM_001300918.1 | NP_001287847.1 | ||
NM_001300919.1 | NP_001287848.1 | |||
NM_003483.4 | NP_003474.1 | |||
NM_003484.1 | NP_003475.1 | |||
XM_017019989.1 | XP_016875478.1 | |||
AB209853.1 | BAD93090.1 | |||
AF533651.1 | ||||
AF533652.1 | ||||
AF533654.1 | AAO48742.1 | |||
AI290477.1 | ||||
AY601861.1 | ||||
AY601862.1 | ||||
AY601863.1 | ||||
AY601864.1 | ||||
AY601865.1 | ||||
AY601866.1 | ||||
AY601867.1 | ||||
CA422738.1 | ||||
U28749.1 | AAA68613.1 | |||
X92518.1 | CAA63295.1 | |||
Z31595.1 | CAA83472.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |