Assay Details
Target Gene Details
Entrez Gene ID: | 5303 |
Gene Name: | peptidylprolyl cis/trans isomerase, NIMA-interacting 4 |
Gene Aliases: |
EPVH, PAR14, PAR17 |
Location: |
Chr.X:72181676-72263964 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIN4 | NM_001170747.1 | NP_001164218.1 | ||
NM_006223.3 | NP_006214.2 | |||
NR_033187.1 | ||||
AB009690.1 | BAA82320.1 | |||
AF143096.1 | AAD27893.1 | |||
AK127605.1 | ||||
AU098526.1 | ||||
AV709926.2 | ||||
BC005234.1 | AAH05234.2 | |||
BC070288.1 | AAH70288.1 | |||
BC093700.1 | AAH93700.1 | |||
BC104653.1 | AAI04654.1 | |||
BC111394.1 | ||||
BC112281.1 | AAI12282.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |