Assay Details
Target Gene Details
Entrez Gene ID: | 1974 |
Gene Name: | eukaryotic translation initiation factor 4A2 |
Gene Aliases: |
BM-010, DDX2B, EIF4A, EIF4F, eIF-4A-II, eIF4A-II |
Location: |
Chr.3:186783572-186789897 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF4A2 | NM_001967.3 | NP_001958.2 | ||
AB209021.1 | 3 | 3192 | BAD92258.1 | |
AF208852.1 | AAF64266.1 | |||
AK130087.1 | ||||
AK225646.1 | ||||
AK296279.1 | 5 | 807 | ||
AL117412.1 | CAI46218.1 | |||
AL137681.1 | 4 | 1838 | ||
AL556981.3 | ||||
BC012547.1 | AAH12547.1 | |||
BC013708.1 | AAH13708.1 | |||
BC015842.1 | AAH15842.1 | |||
BC016295.2 | AAH16295.1 | |||
BC048105.1 | AAH48105.1 | |||
BT009860.1 | AAP88862.1 | |||
D30655.1 | BAA06336.1 | |||
S79942.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893839 | Chr.3:186785646 - 186958667 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2759206 | Chr.3:186619345 - 186849537 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv5005n100 | Chr.3:186709551 - 186836503 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1012615 | Chr.3:186676249 - 186950429 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv34136 | Chr.3:186697095 - 187036942 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)