Assay Details
Target Gene Details
Entrez Gene ID: | 56910 |
Gene Name: | StAR related lipid transfer domain containing 7 |
Gene Aliases: |
GTT1 |
Location: |
Chr.2:96184859-96208846 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STARD7 | NM_020151.3 | NP_064536.2 | ||
XM_011511499.1 | XP_011509801.1 | |||
AF270647.1 | AAF81750.1 | |||
AK023234.1 | ||||
AK054615.1 | ||||
AK291190.1 | ||||
AL542720.3 | ||||
BC007894.2 | AAH07894.1 | |||
BC008894.2 | AAH08894.1 | |||
BC009998.2 | AAH09998.1 | |||
BC012774.2 | AAH12774.1 | |||
BC012793.2 | AAH12793.1 | |||
BC013279.2 | AAH13279.1 | |||
BC014076.2 | AAH14076.1 | |||
BC014274.2 | AAH14274.3 | |||
BC032106.1 | AAH32106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834305 | Chr.2:96183160 - 96353738 on Build GRCh38 | Gain | STARD7 STARD7-AS1 TMEM127 NCAPH LOC105373495 CIAO1 ITPRIPL1 SNRNP200 |
nsv525780 | Chr.2:96193176 - 96193296 on Build GRCh38 | Loss | STARD7 |
dgv607e201 | Chr.2:95855884 - 97561622 on Build GRCh38 | Deletion | LOC100506076 LMAN2L MIR3127 NEURL3 CIAO1 GPAT2 ADRA2B LOC107984110 FAHD2CP ANKRD36B ANKRD36C CNNM3 DUSP2 ANKRD36 STARD7-AS1 TMEM127 ANKRD39 NCAPH KANSL3 LOC105373496 ITPRIPL1 LOC653924 SNRNP200 FER1L5 STARD7 ASTL FAHD2B FAM178B LOC101927053 ARID5A LOC100506123 SEMA4C LOC105373495 CNNM4 ANKRD23 |
nsv1135873 | Chr.2:95981454 - 97579645 on Build GRCh38 | Deletion | LOC100506076 LMAN2L MIR3127 NEURL3 CIAO1 GPAT2 ADRA2B LOC107984110 FAHD2CP ANKRD36B ANKRD36C CNNM3 DUSP2 ANKRD36 STARD7-AS1 TMEM127 ANKRD39 NCAPH KANSL3 LOC105373496 ITPRIPL1 LOC653924 SNRNP200 FER1L5 STARD7 ASTL FAHD2B FAM178B LOC101927053 ARID5A LOC100506123 SEMA4C LOC105373495 CNNM4 ANKRD23 |
esv3425717 | Chr.2:95859900 - 97515905 on Build GRCh38 | Duplication | LOC100506076 LMAN2L MIR3127 NEURL3 CIAO1 GPAT2 ADRA2B LOC107984110 FAHD2CP ANKRD36B ANKRD36C CNNM3 DUSP2 ANKRD36 STARD7-AS1 TMEM127 ANKRD39 NCAPH KANSL3 LOC105373496 ITPRIPL1 LOC653924 SNRNP200 FER1L5 STARD7 ASTL FAHD2B FAM178B LOC101927053 ARID5A LOC100506123 SEMA4C LOC105373495 CNNM4 ANKRD23 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |