Assay Details
Target Gene Details
Entrez Gene ID: | 87 |
Gene Name: | actinin alpha 1 |
Gene Aliases: |
BDPLT15 |
Location: |
Chr.14:68874123-68979366 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACTN1 | NM_001102.3 | 21 | 3427 | NP_001093.1 |
NM_001130004.1 | 22 | 3493 | NP_001123476.1 | |
NM_001130005.1 | 21 | 3412 | NP_001123477.1 | |
XM_011537265.2 | 21 | 3243 | XP_011535567.1 | |
XM_011537266.2 | 21 | 3228 | XP_011535568.1 | |
XM_011537267.2 | 21 | 3219 | XP_011535569.1 | |
XM_011537268.2 | 21 | 3204 | XP_011535570.1 | |
XM_017021720.1 | 21 | 3774 | XP_016877209.1 | |
XM_017021721.1 | 21 | 3750 | XP_016877210.1 | |
XM_017021722.1 | 21 | 3919 | XP_016877211.1 | |
XM_017021723.1 | 21 | 3895 | XP_016877212.1 | |
XM_017021725.1 | 21 | 4093 | XP_016877214.1 | |
XM_017021726.1 | 21 | 4069 | XP_016877215.1 | |
XM_017021727.1 | 21 | 3373 | XP_016877216.1 | |
XM_017021728.1 | 21 | 3523 | XP_016877217.1 | |
AK098203.1 | 14 | 2145 | ||
AK124378.1 | 2 | 2538 | ||
AK294317.1 | ||||
BC015766.1 | 21 | 3234 | AAH15766.1 | |
BI494856.1 | 1 | 245 | ||
BX641076.1 | 10 | 4566 | ||
DQ496098.1 | 22 | 3366 | ||
X55187.1 | 12 | 2229 | CAA38970.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |