Assay Details
Target Gene Details
Entrez Gene ID: | 9329 |
Gene Name: | general transcription factor IIIC subunit 4 |
Gene Aliases: |
KAT12, TF3C-delta, TFIII90, TFIIIC290, TFIIIC90, TFIIICDELTA |
Location: |
Chr.9:132670035-132694955 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GTF3C4 | NM_012204.3 | 2 | 2060 | NP_036336.2 |
NR_133925.1 | 2 | 2060 | ||
XM_017015317.1 | 2 | 1607 | XP_016870806.1 | |
XM_017015318.1 | XP_016870807.1 | |||
AF142328.1 | 2 | 1748 | AAF05087.1 | |
AK024655.1 | 1 | 523 | ||
BC011619.2 | 2 | 1584 | AAH11619.1 | |
BC022787.1 | 2 | 1584 | ||
BC094774.1 | 2 | 1605 | AAH94774.1 | |
BC104755.1 | 2 | 1565 | AAI04756.1 | |
BC112245.1 | 2 | 1565 | AAI12246.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |