Assay Details
Target Gene Details
Entrez Gene ID: | 677 |
Gene Name: | ZFP36 ring finger protein like 1 |
Gene Aliases: |
BRF1, Berg36, ERF-1, ERF1, RNF162B, TIS11B, cMG1 |
Location: |
Chr.14:68787655-68796243 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZFP36L1 | NM_001244701.1 | NP_001231630.1 | ||
DA633985.1 | ||||
FY210721.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1070226 | Chr.14:68788582 - 68795483 on Build GRCh38 | Deletion | ZFP36L1 |
nsv952859 | Chr.14:68788584 - 68795783 on Build GRCh38 | Deletion | ZFP36L1 |
dgv1097n106 | Chr.14:68790783 - 68795883 on Build GRCh38 | Deletion | ZFP36L1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |