Assay Details
Target Gene Details
Entrez Gene ID: | 388428 |
Gene Name: | AATK antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:81165507-81183164 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK-AS1 | NR_027255.1 | 4 | 752 | |
AK126811.1 | 4 | 752 | BAC86704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss | LOC105371925 C17orf89 MIR338 LOC400627 MIR3065 BAIAP2 BAHCC1 HGS FSCN2 LOC107985022 CCDC137 MIR3186 ACTG1 LOC105371921 OXLD1 NPLOC4 LOC107985021 LOC100129503 AATK-AS1 RPTOR LOC101928855 BAIAP2-AS1 CEP131 TMEM105 FAAP100 AATK MIR657 TEPSIN TSPAN10 MRPL12 SLC38A10 LINC00482 LOC100130370 PDE6G SLC25A10 MIR1250 ARL16 MIR4740 MIR6786 CHMP6 |
nsv1057306 | Chr.17:81154954 - 81207813 on Build GRCh38 | Loss | AATK AATK-AS1 CEP131 |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | LOC105371925 C17orf89 ANAPC11 RAC3 GCGR SLC16A3 ZNF750 FN3KRP BAHCC1 PCYT2 HGS LOC105371944 FSCN2 FN3K SNORD134 MIR3186 LOC101929552 MIR4525 AATK-AS1 CCDC57 C17orf62 DCXR NARF CEP131 TMEM105 RFNG TEPSIN MRPL12 SLC38A10 SIRT7 MCRIP1 PDE6G SLC25A10 B3GNTL1 MAFG-AS1 ARL16 SECTM1 CD7 FOXK2 LOC105376790 NPB WDR45B CSNK1D MAFG HEXDC DUS1L TEX19 PYCR1 LOC101929511 PPP1R27 CCDC137 NOTUM ACTG1 OXLD1 NPLOC4 P4HB ALYREF TBCD RAB40B ARHGDIA LRRC45 OGFOD3 ASPSCR1 GPS1 FAAP100 AATK TSPAN10 FASN STRA13 LINC00482 MIR6787 LOC100130370 UTS2R MYADML2 LOC105376791 MIR4740 MIR6786 |
dgv46n68 | Chr.17:81133921 - 81288932 on Build GRCh38 | Loss | AATK LOC105371925 TEPSIN C17orf89 SLC38A10 AATK-AS1 CEP131 |
nsv1065052 | Chr.17:81080876 - 81204920 on Build GRCh38 | Gain | AATK MIR657 MIR338 AATK-AS1 MIR3065 MIR1250 CEP131 BAIAP2 |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | LOC105371925 C17orf89 MIR338 AATK-AS1 MIR3065 BAIAP2-AS1 CEP131 TMEM105 BAIAP2 AATK MIR657 TEPSIN SLC38A10 LINC00482 MIR1250 |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | AATK MIR657 MIR338 AATK-AS1 MIR3065 MIR1250 BAIAP2-AS1 CEP131 BAIAP2 |
nsv1064123 | Chr.17:81168449 - 81282457 on Build GRCh38 | Gain | LOC105371925 TEPSIN C17orf89 SLC38A10 AATK-AS1 CEP131 |
nsv952389 | Chr.17:81137501 - 81593774 on Build GRCh38 | Deletion | NPLOC4 LOC105371925 C17orf89 AATK-AS1 CEP131 TMEM105 BAHCC1 FAAP100 AATK TEPSIN FSCN2 SLC38A10 LINC00482 LOC100130370 MIR4740 MIR3186 ACTG1 |
nsv1066947 | Chr.17:81094837 - 81181290 on Build GRCh38 | Loss | AATK MIR657 MIR338 AATK-AS1 MIR3065 MIR1250 BAIAP2 |