Assay Details
Target Gene Details
Entrez Gene ID: | 7051 |
Gene Name: | transglutaminase 1 |
Gene Aliases: |
ARCI1, ICR2, KTG, LI, LI1, TGASE, TGK |
Location: |
Chr.14:24249114-24263210 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TGM1 | NM_000359.2 | 15 | 2543 | NP_000350.1 |
AK291350.1 | 15 | 2510 | ||
AK299436.1 | 15 | 2312 | ||
AK301652.1 | 9 | 1441 | ||
AK315819.1 | 15 | 2543 | ||
AK315828.1 | 15 | 2503 | ||
AK315843.1 | 15 | 2510 | ||
BC034699.1 | 15 | 2494 | AAH34699.1 | |
D90287.1 | 15 | 2499 | BAA14329.1 | |
M55183.1 | 15 | 2472 | AAA59474.1 | |
M62925.1 | 15 | 2505 | AAA61166.1 | |
X57974.1 | 9 | 1297 | CAA41040.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |