Assay Details
Target Gene Details
Entrez Gene ID: | 11274 |
Gene Name: | ubiquitin specific peptidase 18 |
Gene Aliases: |
ISG43, UBP43 |
Location: |
Chr.22:18149959-18177397 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP18 | NM_017414.3 | NP_059110.2 | ||
XM_006724074.2 | XP_006724137.1 | |||
AI222587.1 | ||||
AJ243526.1 | CAB76398.1 | |||
AK313385.1 | ||||
AL136690.1 | CAB66625.1 | |||
BC014896.1 | AAH14896.1 | |||
BI752849.1 | ||||
CT841507.1 | CAJ86437.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv173e55 | Chr.22:18157533 - 19070277 on Build GRCh38 | Gain | LOC100996415 PRODH TMEM191B LOC729461 PI4KAP1 DGCR11 RIMBP3 DGCR10 GGT3P LOC100996432 DGCR6 USP18 DGCR5 LOC642643 DGCR2 LOC100996401 FAM230A DGCR9 LOC102725072 |
nsv516997 | Chr.22:18143870 - 19039100 on Build GRCh38 | Gain+Loss | LOC100996415 PRODH TMEM191B LOC729461 PI4KAP1 RIMBP3 DGCR10 GGT3P LOC100996432 DGCR6 USP18 DGCR5 LOC642643 DGCR2 LOC100996401 FAM230A DGCR9 LOC102725072 |
nsv1055756 | Chr.22:18062621 - 18794021 on Build GRCh38 | Gain | LOC100996415 TMEM191B USP18 LOC729461 LOC642643 LOC100996401 PI4KAP1 PEX26 RIMBP3 TUBA8 FAM230A GGT3P |
nsv1058565 | Chr.22:18060494 - 18159378 on Build GRCh38 | Gain | USP18 PEX26 TUBA8 |
esv2751935 | Chr.22:18149609 - 19042823 on Build GRCh38 | Loss | LOC100996415 PRODH TMEM191B LOC729461 PI4KAP1 RIMBP3 DGCR10 GGT3P LOC100996432 DGCR6 USP18 DGCR5 LOC642643 DGCR2 LOC100996401 FAM230A DGCR9 LOC102725072 |
esv3893430 | Chr.22:18059545 - 18169093 on Build GRCh38 | Gain | USP18 PEX26 TUBA8 |
esv2758824 | Chr.22:18069029 - 19074200 on Build GRCh38 | Gain+Loss | LOC100996415 PRODH TMEM191B LOC729461 PI4KAP1 PEX26 DGCR11 RIMBP3 TUBA8 DGCR10 GGT3P LOC100996432 DGCR6 USP18 DGCR5 LOC642643 DGCR2 LOC100996401 FAM230A DGCR9 LOC102725072 |
nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss | LOC100996415 TMEM191B ATP6V1E1 LINC00528 TUBA8 GGT3P LOC100996432 SLC25A18 LOC100996401 MIR648 BCL2L13 LOC729461 PI4KAP1 PEX26 RIMBP3 MICAL3 FLJ41941 USP18 LOC642643 LOC101929372 BID MIR3198-1 FAM230A CECR2 LOC102725072 |
nsv471594 | Chr.22:18076012 - 18882129 on Build GRCh38 | Gain+Loss | LOC100996415 TMEM191B LOC729461 PI4KAP1 PEX26 RIMBP3 TUBA8 GGT3P LOC100996432 USP18 LOC642643 LOC100996401 FAM230A LOC102725072 |
nsv469560 | Chr.22:18076011 - 18213970 on Build GRCh38 | Gain | LOC100996415 USP18 PEX26 TUBA8 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |