Assay Details
Target Gene Details
Entrez Gene ID: | 388428 |
Gene Name: | AATK antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:81165507-81183164 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AATK-AS1 | NR_027255.1 | |||
AK126811.1 | BAC86704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv828128 | Chr.17:80659041 - 81736487 on Build GRCh38 | Loss | LOC400627 BAHCC1 BAIAP2 MIR3065 PDE6G TSPAN10 MIR3186 NPLOC4 AATK RPTOR BAIAP2-AS1 ACTG1 MIR1250 CEP131 MRPL12 AATK-AS1 FAAP100 LOC107985021 HGS CCDC137 LOC101928855 LOC100130370 OXLD1 TEPSIN TMEM105 LINC00482 SLC25A10 MIR657 LOC107985022 MIR6786 FSCN2 ARL16 SLC38A10 CHMP6 LOC100129503 C17orf89 LOC105371925 LOC105371921 MIR4740 MIR338 |
nsv1057306 | Chr.17:81154954 - 81207813 on Build GRCh38 | Loss | CEP131 AATK-AS1 AATK |
nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain | ASPSCR1 NPB BAHCC1 CCDC57 TSPAN10 B3GNTL1 TEX19 SIRT7 LOC105376790 ACTG1 LOC101929511 TBCD FOXK2 C17orf62 MYADML2 LOC105371944 SECTM1 MCRIP1 PPP1R27 WDR45B LOC100130370 PCYT2 TEPSIN TMEM105 SLC25A10 GCGR ANAPC11 ARL16 FASN ARHGDIA SLC16A3 MIR4740 FN3KRP FN3K P4HB RFNG RAC3 PDE6G MIR3186 NPLOC4 AATK RAB40B CEP131 NOTUM STRA13 MRPL12 UTS2R GPS1 AATK-AS1 CSNK1D OGFOD3 MIR4525 MIR6787 FAAP100 CD7 NARF DCXR HGS ALYREF SNORD134 CCDC137 HEXDC MAFG OXLD1 MAFG-AS1 LRRC45 LINC00482 MIR6786 ZNF750 FSCN2 LOC105376791 SLC38A10 C17orf89 LOC105371925 LOC101929552 PYCR1 DUS1L |
dgv46n68 | Chr.17:81133921 - 81288932 on Build GRCh38 | Loss | CEP131 SLC38A10 C17orf89 LOC105371925 AATK-AS1 TEPSIN AATK |
nsv1065052 | Chr.17:81080876 - 81204920 on Build GRCh38 | Gain | MIR657 BAIAP2 MIR1250 CEP131 MIR3065 AATK-AS1 MIR338 AATK |
nsv470619 | Chr.17:81032233 - 81349843 on Build GRCh38 | Loss | BAIAP2 MIR3065 TEPSIN AATK TMEM105 LINC00482 MIR657 BAIAP2-AS1 MIR1250 CEP131 SLC38A10 C17orf89 LOC105371925 AATK-AS1 MIR338 |
dgv3292n100 | Chr.17:81019582 - 81204920 on Build GRCh38 | Gain | MIR657 BAIAP2-AS1 BAIAP2 MIR1250 CEP131 MIR3065 AATK-AS1 MIR338 AATK |
nsv1064123 | Chr.17:81168449 - 81282457 on Build GRCh38 | Gain | CEP131 SLC38A10 C17orf89 LOC105371925 AATK-AS1 TEPSIN |
nsv952389 | Chr.17:81137501 - 81593774 on Build GRCh38 | Deletion | BAHCC1 LOC100130370 TEPSIN MIR3186 NPLOC4 AATK TMEM105 LINC00482 ACTG1 FSCN2 CEP131 SLC38A10 C17orf89 LOC105371925 AATK-AS1 MIR4740 FAAP100 |