Assay Details
Target Gene Details
Entrez Gene ID: | 25897 |
Gene Name: | ring finger protein 19A, RBR E3 ubiquitin protein ligase |
Gene Aliases: |
RNF19 |
Location: |
Chr.8:100257059-100336218 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RNF19A | NM_001280539.1 | 4 | 1117 | NP_001267468.1 |
NM_015435.4 | 4 | 1148 | NP_056250.3 | |
NM_183419.3 | 3 | 1224 | NP_904355.1 | |
XM_005250853.3 | 3 | 1186 | XP_005250910.1 | |
XM_011516956.2 | 3 | 1209 | XP_011515258.1 | |
XM_017013302.1 | 4 | 1309 | XP_016868791.1 | |
XM_017013303.1 | 4 | 1219 | XP_016868792.1 | |
XM_017013304.1 | 3 | 1166 | XP_016868793.1 | |
XM_017013305.1 | XP_016868794.1 | |||
AB029316.1 | 4 | 1148 | BAB39353.1 | |
BC093938.1 | 2 | 920 | AAH93938.1 | |
BC093940.2 | 2 | 920 | AAH93940.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |