Assay Details
Target Gene Details
Entrez Gene ID: | 79901 |
Gene Name: | cytochrome b reductase 1 |
Gene Aliases: |
CYB561A2, DCYTB, FRRS3 |
Location: |
Chr.2:171522247-171558133 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYBRD1 | NM_001127383.1 | 1 | 282 | NP_001120855.1 |
NM_001256909.1 | NP_001243838.1 | |||
NM_024843.3 | 1 | 282 | NP_079119.3 | |
AK027115.1 | 1 | 124 | BAB15661.1 | |
AK128495.1 | 1 | 161 | BAC87466.1 | |
AK225459.1 | 1 | 124 | ||
AK291731.1 | 1 | 132 | ||
AK301485.1 | ||||
AK316588.1 | 1 | 161 | ||
AL136693.1 | 1 | 165 | CAB66628.1 | |
BC065290.1 | 1 | 117 | AAH65290.1 | |
CB266594.1 | 1 | 265 | ||
CX866825.1 | 1 | 173 | ||
DA211209.1 | 1 | 405 | ||
DB235339.1 | 1 | 282 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |