Assay Details
Target Gene Details
Entrez Gene ID: | 23493 |
Gene Name: | hes related family bHLH transcription factor with YRPW motif 2 |
Gene Aliases: |
CHF1, GRIDLOCK, GRL, HERP1, HESR2, HRT2, bHLHb32 |
Location: |
Chr.6:125747592-125762243 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HEY2 | NM_012259.2 | 5 | 723 | NP_036391.1 |
XM_017010627.1 | 5 | 2107 | XP_016866116.1 | |
XM_017010628.1 | 6 | 1805 | XP_016866117.1 | |
XM_017010629.1 | 5 | 584 | XP_016866118.1 | |
AB044755.1 | 5 | 628 | BAA96781.1 | |
AF173901.1 | 5 | 691 | AAF20173.1 | |
AF232238.1 | 5 | 642 | AAF37296.1 | |
AF237949.1 | 5 | 599 | AAF44781.1 | |
AF311884.1 | 5 | 532 | AAG31157.1 | |
AJ249545.1 | 5 | 582 | CAB56839.1 | |
AK289943.1 | 5 | 641 | ||
AK303890.1 | 5 | 545 | ||
BC007707.2 | 5 | 621 | AAH07707.1 | |
BT020067.1 | 5 | 532 | AAV38870.1 | |
BT020068.1 | 5 | 532 | AAV38871.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |