Assay Details
Target Gene Details
Entrez Gene ID: | 160 |
Gene Name: | adaptor related protein complex 2 alpha 1 subunit |
Gene Aliases: |
ADTAA, AP2-ALPHA, CLAPA1 |
Location: |
Chr.19:49766923-49807114 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AP2A1 | NM_014203.2 | NP_055018.2 | ||
NM_130787.2 | NP_570603.2 | |||
XM_011526556.2 | XP_011524858.1 | |||
XM_011526557.2 | XP_011524859.1 | |||
AK124343.1 | ||||
AK302005.1 | ||||
AL136925.1 | CAB66859.1 | |||
BC014214.2 | AAH14214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | GFY PLEKHA4 GRWD1 NUCB1 MIR150 C19orf73 ZNF473 TMEM143 TSKS FLT3LG PLA2G4C SNAR-A7 ALDH16A1 MIR4324 KDELR1 LOC101059948 MIR5088 GRIN2D SULT2B1 CGB8 SLC6A16 LHB TULP2 MIR4749 CPT1C RPL18 RPL13A SNAR-A1 PTOV1-AS2 ELSPBP1 SIGLEC16 FCGRT AKT1S1 SNAR-G2 SNAR-A12 HSD17B14 SNAR-C1 BAX ATF5 SNAR-C2 PRMT1 FUT1 DBP MIR6799 MIR4751 CA11 SNAR-A6 CGB3 BSPH1 MAMSTR VRK3 CARD8-AS1 CGB7 CYTH2 CGB5 SNAR-A10 NUCB1-AS1 PLA2G4C-AS1 SNAR-D SNAR-C5 SNAR-A9 FGF21 SNAR-A3 TBC1D17 HRC RRAS RASIP1 SYNGR4 PRR12 NTF4 SNAR-A13 FTL ADM5 SNAR-A14 FUZ TEAD2 SIGLEC11 SNORD35B SLC17A7 CABP5 PRRG2 LIG1 IRF3 SNAR-A2 MIR6800 SNAR-C4 LOC105447645 PTH2 RUVBL2 PNKP KCNA7 LOC101928295 IZUMO1 SEC1P SNORD35A PTOV1 BCL2L12 SNAR-C3 ZNF114 MIR6798 NUP62 CGB1 DHDH SNAR-B2 RPS11 MIR4750 SNAR-G1 KCNJ14 SPACA4 LOC105372430 PTOV1-AS1 RCN3 FUT2 PPP1R15A PIH1D1 TRPM4 NOSIP SNAR-A11 CARD8 SNAR-A5 IL4I1 SNAR-A4 SNORD32A CD37 SNORD34 MED25 NTN5 AP2A1 EMP3 SNRNP70 CGB2 SNAR-B1 PPFIA3 BCAT2 SNAR-A8 LOC100287477 FLJ26850 LMTK3 C19orf68 SCAF1 GYS1 CCDC155 LIN7B SPHK2 CCDC114 SNORD33 FAM83E DKKL1 |
nsv470150 | Chr.19:49618821 - 49856789 on Build GRCh38 | Gain | MIR5088 PTOV1-AS1 FUZ PRMT1 MIR6799 MIR4749 CPT1C PTOV1 BCL2L12 TSKS RRAS SCAF1 PRR12 MED25 PTOV1-AS2 IRF3 AP2A1 MIR6800 ADM5 |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss | PTOV1-AS1 FUZ ATF5 PNKP MIR6799 MIR4751 MIR4749 PTOV1 TSKS IL4I1 TBC1D17 MED25 PTOV1-AS2 NUP62 AP2A1 MIR4750 MIR6800 AKT1S1 |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | PTOV1-AS1 FUZ ATF5 PRMT1 NOSIP MIR6799 MIR4751 TSKS IL4I1 PRRG2 MED25 IRF3 AP2A1 MIR6800 MIR5088 PNKP MIR4749 CPT1C PTOV1 BCL2L12 TBC1D17 RRAS SCAF1 PRR12 PTOV1-AS2 NUP62 MIR4750 ADM5 AKT1S1 |
nsv1160655 | Chr.19:49489110 - 49841830 on Build GRCh38 | Deletion | PTOV1-AS1 RCN3 FUZ MIR150 PRMT1 NOSIP MIR6799 TSKS SNORD32A SNORD35B SNORD34 PRRG2 MED25 IRF3 AP2A1 MIR6800 MIR5088 CPT1C SNORD35A BCL2L12 RRAS SCAF1 RPL13A PRR12 SNORD33 RPS11 FCGRT ADM5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |