Assay Details
Target Gene Details
Entrez Gene ID: | 83855 |
Gene Name: | Kruppel like factor 16 |
Gene Aliases: |
BTEB4, DRRF, NSLP2 |
Location: |
Chr.19:1852399-1863565 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLF16 | NM_031918.3 | 2 | 2648 | NP_114124.1 |
AF327440.1 | AAK15698.1 | |||
AK025941.1 | 1 | 1573 | ||
BQ447922.1 | 1 | 257 | ||
BU594659.1 | 1 | 255 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893156 | Chr.19:1766738 - 2115791 on Build GRCh38 | Loss | IZUMO4 ONECUT3 MKNK2 SCAMP4 AP3D1 ATP8B3 ABHD17A CSNK1G2 ADAT3 CSNK1G2-AS1 KLF16 LOC100288123 MOB3A BTBD2 REXO1 MIR1909 |
esv2669985 | Chr.19:1852046 - 1864993 on Build GRCh38 | Deletion | KLF16 |
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | MEX3D ONECUT3 SCAMP4 APC2 ATP8B3 ABHD17A TCF3 ADAT3 CSNK1G2-AS1 BTBD2 MBD3 DAZAP1 MUM1 UQCR11 MIR1909 PCSK4 C19orf25 REEP6 GAMT RPS15 CSNK1G2 NDUFS7 PLK5 KLF16 LOC100288123 REXO1 ADAMTSL5 |
nsv470107 | Chr.19:1782799 - 2100347 on Build GRCh38 | Loss | IZUMO4 MKNK2 SCAMP4 ATP8B3 ABHD17A CSNK1G2 ADAT3 CSNK1G2-AS1 KLF16 LOC100288123 MOB3A BTBD2 REXO1 MIR1909 |
nsv1160560 | Chr.19:1791100 - 1916393 on Build GRCh38 | Deletion | LOC100288123 SCAMP4 ATP8B3 ABHD17A REXO1 MIR1909 ADAT3 KLF16 |
nsv578338 | Chr.19:1847699 - 1869095 on Build GRCh38 | Loss | REXO1 KLF16 |
nsv578337 | Chr.19:1808864 - 1904613 on Build GRCh38 | Gain | LOC100288123 ATP8B3 ABHD17A REXO1 MIR1909 KLF16 |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | MEX3D ONECUT3 MKNK2 SCAMP4 APC2 ATP8B3 ABHD17A TCF3 ADAT3 CSNK1G2-AS1 BTBD2 MBD3 DAZAP1 MUM1 UQCR11 MIR1909 PCSK4 C19orf25 REEP6 GAMT RPS15 CSNK1G2 NDUFS7 PLK5 KLF16 LOC100288123 REXO1 ADAMTSL5 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |