Assay Details
Target Gene Details
Entrez Gene ID: | 56985 |
Gene Name: | ADP-ribose/CDP-alcohol diphosphatase, manganese dependent |
Gene Aliases: |
C17orf48, MDS006, NBLA03831 |
Location: |
Chr.17:10697610-10711558 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADPRM | NM_020233.4 | 4 | 850 | NP_064618.3 |
XM_005256738.2 | 4 | 846 | XP_005256795.1 | |
AB073393.1 | 4 | 821 | BAE45723.1 | |
KF880964.1 | 3 | 775 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2675560 | Chr.17:10710661 - 10748164 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() |
nsv1065819 | Chr.17:10622619 - 10784575 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3639941 | Chr.17:10710674 - 10748133 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv3639942 | Chr.17:10710767 - 10713237 on Build GRCh38 | Loss |
![]() ![]() |
nsv1063900 | Chr.17:10662646 - 10729182 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv833367 | Chr.17:10683841 - 10826633 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1058835 | Chr.17:10322963 - 10939602 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)