Assay Details
Target Gene Details
Entrez Gene ID: | 57407 |
Gene Name: | NmrA-like family domain containing 1 |
Gene Aliases: |
HSCARG, SDR48A1 |
Location: |
Chr.16:4461677-4476320 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NMRAL1 | NM_001305141.1 | 6 | 1227 | NP_001292070.1 |
NM_001305142.1 | 6 | 1214 | NP_001292071.1 | |
NM_020677.4 | 6 | 1328 | NP_065728.1 | |
XM_006720905.3 | XP_006720968.1 | |||
XM_006720906.2 | XP_006720969.1 | |||
XM_017023485.1 | XP_016878974.1 | |||
XM_017023489.1 | XP_016878978.1 | |||
XM_017023490.1 | XP_016878979.1 | |||
XM_017023491.1 | XP_016878980.1 | |||
XM_017023492.1 | XP_016878981.1 | |||
AF225419.1 | AAG09721.1 | |||
AK098784.1 | ||||
AW246763.1 | 1 | 58 | ||
BC002927.2 | AAH02927.1 | |||
BC007364.2 | AAH07364.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160332 | Chr.16:4367643 - 4486554 on Build GRCh38 | Duplication |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv833132 | Chr.16:4370092 - 4463448 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3584924 | Chr.16:4330628 - 4469258 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv23732 | Chr.16:4340813 - 4476244 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)