Assay Details
Target Gene Details
Entrez Gene ID: | 80194 |
Gene Name: | transmembrane protein 134 |
Gene Aliases: |
- |
Location: |
Chr.11:67464348-67469277 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM134 | NM_001078650.2 | 6 | 631 | NP_001072118.1 |
NM_001078651.2 | 7 | 649 | NP_001072119.1 | |
NM_025124.3 | 7 | 676 | NP_079400.1 | |
NR_073409.1 | 7 | 843 | ||
NR_073410.1 | 6 | 615 | ||
NR_073411.1 | 7 | 687 | ||
NR_073412.1 | 7 | 808 | ||
NR_073413.1 | 7 | 660 | ||
XM_006718693.2 | 6 | 599 | XP_006718756.1 | |
XM_017018356.1 | 6 | 777 | XP_016873845.1 | |
AA830850.1 | 1 | 307 | ||
AK025402.1 | 7 | 692 | BAB15126.1 | |
AW249982.1 | 1 | 272 | ||
BC125134.1 | 6 | 599 | ||
BG488634.1 | 7 | 598 | ||
BG719036.1 | 6 | 611 | ||
BX400047.2 | 7 | 764 | ||
CN336445.1 | 7 | 510 | ||
DN999439.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv468604 | Chr.11:67418529 - 67516420 on Build GRCh38 | Loss | CARNS1 CORO1B AIP PITPNM1 RPS6KB2 MIR6752 CDK2AP2 GPR152 TMEM134 CABP4 PTPRCAP |
dgv1977n54 | Chr.11:67397024 - 67521123 on Build GRCh38 | Loss | RAD9A AIP PITPNM1 CDK2AP2 GPR152 TMEM134 PPP1CA CARNS1 CORO1B CABP2 TBC1D10C RPS6KB2 MIR6752 CABP4 PTPRCAP |
dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion | RAD9A AIP PITPNM1 CLCF1 CDK2AP2 GPR152 TMEM134 PPP1CA LOC100130987 CARNS1 CORO1B CABP2 TBC1D10C RPS6KB2 MIR6752 CABP4 PTPRCAP |
nsv521723 | Chr.11:67408221 - 67543787 on Build GRCh38 | Loss | AIP PITPNM1 CDK2AP2 GPR152 TMEM134 CARNS1 CORO1B CABP2 TBC1D10C RPS6KB2 MIR6752 CABP4 PTPRCAP |
nsv527056 | Chr.11:67439955 - 67526837 on Build GRCh38 | Loss | CORO1B AIP CABP2 PITPNM1 MIR6752 CDK2AP2 GPR152 TMEM134 CABP4 |