Assay Details
Target Gene Details
Entrez Gene ID: | 9585 |
Gene Name: | kinesin family member 20B |
Gene Aliases: |
CT90, KRMP1, MPHOSPH1, MPP-1, MPP1 |
Location: |
Chr.10:89701590-89774943 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KIF20B | NM_001284259.1 | 2 | 148 | NP_001271188.1 |
NM_016195.3 | 2 | 148 | NP_057279.2 | |
AB033337.1 | 2 | 101 | BAB69456.1 | |
AK074336.1 | 2 | 99 | ||
AK290439.1 | 2 | 128 | ||
AL117496.1 | 2 | 103 | CAB55962.1 | |
AY282406.1 | 2 | 125 | AAP40330.1 | |
BC046134.1 | 2 | 101 | AAH46134.1 | |
BC058913.1 | 2 | 104 | AAH58913.1 | |
BC093089.1 | 2 | 103 | AAH93089.1 | |
BC108688.1 | 2 | 110 | AAI08689.1 | |
CB130618.1 | 2 | 148 |
More Information
Additional Information:
For this assay, SNP(s) [rs114316057] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |