Assay Details
Target Gene Details
Entrez Gene ID: | 3425 |
Gene Name: | iduronidase, alpha-L- |
Gene Aliases: |
IDA, MPS1 |
Location: |
Chr.4:986997-1004557 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IDUA | NM_000203.4 | NP_000194.2 | ||
NR_110313.1 | ||||
XM_017008163.1 | XP_016863652.1 | |||
AB208901.1 | 3 | 961 | BAD92138.1 | |
AK125223.1 | ||||
AK291816.1 | ||||
BC029959.1 | AAH29959.1 | |||
DA057148.1 | ||||
M74715.1 | AAA81589.1 |
Target Gene Details
Entrez Gene ID: | 10861 |
Gene Name: | solute carrier family 26 member 1 |
Gene Aliases: |
CAON, EDM4, SAT-1, SAT1 |
Location: |
Chr.4:979073-993440 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC26A1 | NM_022042.3 | 3 | 1469 | NP_071325.2 |
NM_134425.2 | NP_602297.1 | |||
NM_213613.3 | 4 | 1684 | NP_998778.1 | |
AF297659.2 | 3 | 1424 | AAG22075.1 | |
AK292747.1 | 3 | 1469 | ||
AY124771.1 | 4 | 1680 | AAM94171.1 | |
BC015517.2 | AAH15517.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv482086 | Chr.4:987659 - 993436 on Build GRCh38 | Gain |
![]() ![]() |
nsv822428 | Chr.4:968896 - 1028144 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv1003219 | Chr.4:776106 - 1111676 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3599384 | Chr.4:974637 - 1010875 on Build GRCh38 | Gain |
![]() ![]() |
nsv593226 | Chr.4:948218 - 1025223 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv4196 | Chr.4:961755 - 1004599 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv950328 | Chr.4:988213 - 1101612 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() |
nsv1078530 | Chr.4:987811 - 994112 on Build GRCh38 | Duplication |
![]() ![]() |
nsv469998 | Chr.4:815805 - 1093743 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1008352 | Chr.4:819605 - 1041628 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)