Assay Details
Target Gene Details
Entrez Gene ID: | 11101 |
Gene Name: | arginyltransferase 1 |
Gene Aliases: |
- |
Location: |
Chr.10:121740421-121928801 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATE1 | NM_001001976.2 | 12 | 1709 | NP_001001976.1 |
NM_001288734.1 | 11 | 1605 | NP_001275663.1 | |
NM_001288735.1 | 10 | 1582 | NP_001275664.1 | |
NM_001288736.1 | 12 | 1944 | NP_001275665.1 | |
NM_007041.3 | 12 | 1709 | NP_008972.2 | |
NR_110006.1 | 10 | 1651 | ||
XM_005269458.2 | 13 | 1850 | XP_005269515.1 | |
XM_005269459.3 | 13 | 2073 | XP_005269516.1 | |
XM_017015597.1 | 10 | 1519 | XP_016871086.1 | |
XM_017015598.1 | 12 | 1928 | XP_016871087.1 | |
XM_017015599.1 | 11 | 1592 | XP_016871088.1 | |
XM_017015600.1 | 12 | 1572 | XP_016871089.1 | |
XM_017015602.1 | 11 | 1489 | XP_016871091.1 | |
XM_017015604.1 | 10 | 1434 | XP_016871093.1 | |
AF079098.1 | 12 | 1583 | AAD12366.1 | |
AF079099.1 | 12 | 1583 | AAD12367.1 | |
AK124669.1 | 10 | 1659 | ||
AK292657.1 | 7 | 825 | ||
AK295642.1 | 7 | 941 | ||
AK296356.1 | 11 | 1443 | ||
AK299101.1 | 12 | 1595 | ||
AK303604.1 | 10 | 1540 | ||
AK308926.1 | 5 | 738 | ||
AK313057.1 | ||||
BC022026.1 | 12 | 1682 | AAH22026.2 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |