Assay Details
Target Gene Details
Entrez Gene ID: | 9702 |
Gene Name: | centrosomal protein 57 |
Gene Aliases: |
MVA2, PIG8, TSP57 |
Location: |
Chr.11:95790461-95832693 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CEP57 | NM_001243776.1 | NP_001230705.1 | ||
NM_001243777.1 | NP_001230706.1 | |||
NM_014679.4 | NP_055494.2 | |||
XM_006718945.2 | XP_006719008.1 | |||
XM_006718946.2 | XP_006719009.1 | |||
XM_017018592.1 | XP_016874081.1 | |||
XM_017018593.1 | XP_016874082.1 | |||
XM_017018594.1 | XP_016874083.1 | |||
AK098647.1 | ||||
AK291245.1 | ||||
AK293277.1 | ||||
AY225092.1 | AAO73938.1 | |||
AY239292.1 | AAP72184.1 | |||
BC001233.2 | AAH01233.1 | |||
BC029385.1 | ||||
BC039711.1 | AAH39711.1 | |||
BG699097.1 | ||||
D42054.1 | ||||
DB032301.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832235 | Chr.11:95753612 - 95935071 on Build GRCh38 | Gain | FAM76B CEP57 MTMR2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |