Assay Details
Target Gene Details
Entrez Gene ID: | 57658 |
Gene Name: | calcium binding and coiled-coil domain 1 |
Gene Aliases: |
Cocoa, PP13275, calphoglin |
Location: |
Chr.12:53711118-53727523 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CALCOCO1 | NM_001143682.1 | 8 | 1020 | NP_001137154.1 |
NM_020898.2 | 9 | 1275 | NP_065949.1 | |
NR_026554.1 | 8 | 1172 | ||
XM_011538601.1 | 9 | 1209 | XP_011536903.1 | |
XM_011538602.1 | 10 | 1342 | XP_011536904.1 | |
XM_011538603.1 | 10 | 1360 | XP_011536905.1 | |
XM_011538604.1 | 10 | 1316 | XP_011536906.1 | |
XM_017019707.1 | 10 | 1342 | XP_016875196.1 | |
XM_017019708.1 | 8 | 1337 | XP_016875197.1 | |
XM_017019709.1 | 8 | 1337 | XP_016875198.1 | |
AB040969.1 | 9 | 1172 | BAA96060.1 | |
AF370415.1 | 9 | 1180 | AAQ15251.1 | |
AK027881.1 | 9 | 1209 | BAB55428.1 | |
AK092788.1 | 8 | 1019 | ||
AK122773.1 | 8 | 936 | ||
AK222646.1 | ||||
AK294424.1 | 9 | 1183 | ||
AK296497.1 | 8 | 1103 | ||
AK315578.1 | 9 | 1210 | ||
AL136895.1 | 9 | 1187 | CAB66829.1 | |
AY211909.1 | 5 | 694 | AAO65163.1 | |
AY358397.1 | 9 | 1184 | AAQ88763.1 | |
AY563137.1 | 9 | 1182 | AAT68474.1 | |
BC003177.1 | 9 | 1182 | AAH03177.1 | |
CR533567.1 | 8 | 1131 | CAG38598.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |