Assay Details
Target Gene Details
Entrez Gene ID: | 57611 |
Gene Name: | immunoglobulin superfamily containing leucine rich repeat 2 |
Gene Aliases: |
LINX |
Location: |
Chr.15:74128203-74141826 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ISLR2 | NM_001130136.1 | 4 | 3289 | NP_001123608.1 |
NM_001130137.1 | 4 | 3025 | NP_001123609.1 | |
NM_001130138.1 | 4 | 2733 | NP_001123610.1 | |
NM_020851.2 | 3 | 2858 | NP_065902.1 | |
XM_011521840.2 | 4 | 3128 | XP_011520142.1 | |
XM_011521841.1 | 4 | 2811 | XP_011520143.1 | |
XM_017022446.1 | XP_016877935.1 | |||
AB040898.2 | 4 | 3289 | ||
AK092297.1 | 3 | 2684 | ||
AK092302.1 | ||||
AK096602.1 | 2 | 2636 | ||
AY358316.1 | 3 | 2656 | AAQ88682.1 | |
BC114551.1 | 3 | 1372 | AAI14552.1 | |
BC152429.1 | 4 | 3289 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833053 | Chr.15:74067815 - 74262799 on Build GRCh38 | Loss | STRA6 ISLR2 LOC283731 CCDC33 GOLGA6A LOC732265 ISLR |
nsv1050884 | Chr.15:74102642 - 74197072 on Build GRCh38 | Gain | STRA6 ISLR2 LOC283731 ISLR |
nsv469619 | Chr.15:73999518 - 74173980 on Build GRCh38 | Loss | ISLR2 LOC283731 GOLGA6A PML LOC732265 ISLR |
nsv827385 | Chr.15:74127198 - 74136960 on Build GRCh38 | Gain | ISLR2 LOC283731 |
nsv951993 | Chr.15:74124460 - 74137359 on Build GRCh38 | Deletion | ISLR2 LOC283731 |
esv2760040 | Chr.15:73922773 - 74172575 on Build GRCh38 | Gain+Loss | LOXL1-AS1 ISLR2 LOC283731 GOLGA6A LOXL1 PML LOC732265 STOML1 |
nsv1047564 | Chr.15:74116495 - 74165840 on Build GRCh38 | Gain | ISLR2 LOC283731 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | STRA6 C15orf39 CYP1A2 GOLGA6A FAM219B RPP25 LOC732265 MIR4513 LMAN1L C15orf59-AS1 GOLGA6C UBL7-AS1 CYP1A1 CSK HIGD2B REC114 EDC3 ULK3 LOC107984728 HCN4 UBL7 CYP11A1 LOXL1-AS1 PPCDC BBS4 LOC105376731 CD276 STOML1 LOC102723750 LOXL1 SCAMP5 PML SCAMP2 NEO1 CLK3 LOC283731 CCDC33 TBC1D21 NPTN-IT1 COX5A NPTN ADPGK-AS1 MIR6882 LOC101929333 ISLR ARID3B MIR6881 ISLR2 MPI ADPGK LOC729739 CPLX3 C15orf59 SEMA7A |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |