Assay Details
Target Gene Details
Entrez Gene ID: | 8622 |
Gene Name: | phosphodiesterase 8B |
Gene Aliases: |
ADSD, PPNAD3 |
Location: |
Chr.5:77180252-77428256 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
Target Gene Details
Entrez Gene ID: | 55255 |
Gene Name: | WD repeat domain 41 |
Gene Aliases: |
MSTP048 |
Location: |
Chr.5:77426652-77620611 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WDR41 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic non-DGV Variation |