Assay Details
Target Gene Details
Entrez Gene ID: | 28972 |
Gene Name: | signal peptidase complex subunit 1 |
Gene Aliases: |
HSPC033, SPC1, SPC12, YJR010C-A |
Location: |
Chr.3:52705841-52708182 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPCS1 | NM_014041.3 | 4 | 785 | NP_054760.3 |
AF092138.1 | 4 | 514 | AAD40380.1 | |
AK027426.1 | 4 | 514 | ||
AX775783.1 | 4 | 514 | CAE11591.1 | |
AX775785.1 | 5 | 1299 | CAE11592.1 | |
AX775787.1 | 4 | 494 | CAE11593.1 | |
AX775833.1 | 4 | 504 | CAE11616.1 | |
BC000884.1 | 4 | 494 | AAH00884.1 | |
BI826431.1 | 4 | 479 | ||
BQ423345.1 | 4 | 521 | ||
CA309210.1 | 1 | 316 | ||
L38852.1 | 4 | 504 | AAL31361.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508219 | Chr.3:52707177 - 52769883 on Build GRCh38 | Deletion | SPCS1 NEK4 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |