Assay Details
Target Gene Details
Entrez Gene ID: | 1621 |
Gene Name: | dopamine beta-hydroxylase |
Gene Aliases: |
DBM |
Location: |
Chr.9:133636363-133659344 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DBH | NM_000787.3 | 1 | 222 | NP_000778.3 |
BC017174.2 | 1 | 208 | AAH17174.1 | |
CD014125.1 | ||||
X13255.1 | 1 | 203 | CAA31631.1 | |
X13256.1 | 1 | 203 | CAA31632.1 | |
Y00096.1 | 1 | 210 | CAA68285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion | PPP1R26-AS1 SPACA9 SURF2 OBP2B COL5A1-AS1 LCN1 SNORD36B MRPS2 SURF4 LOC401557 CEL LOC101928193 MIR3689E TMEM8C ADAMTS13 ABO SURF6 AK8 RALGDS COL5A1 MIR3689C MIR3689F DBH-AS1 RNU6ATAC GFI1B LOC101448202 RPL7A GTF3C5 MIR3689B WDR5 SARDH MIR3689D2 OLFM1 PPP1R26 MED22 STKLD1 VAV2 MIR4669 FCN2 C9orf116 LOC100996574 CELP ADAMTSL2 SURF1 TSC1 MIR6877 MIR548AW RXRA FCN1 BRD3 DBH C9orf62 SLC2A6 LOC105376306 MIR3689A CACFD1 SNORD24 SNORD36A LOC101928525 SNORD141A MIR3689D1 FAM163B LINC00094 GBGT1 REXO4 SNORD36C LOC100130548 |
nsv615638 | Chr.9:133633021 - 133640119 on Build GRCh38 | Loss | DBH |
nsv951800 | Chr.9:133627979 - 133696678 on Build GRCh38 | Deletion | DBH-AS1 SARDH DBH |
nsv469906 | Chr.9:133629444 - 133643961 on Build GRCh38 | Loss | DBH |
esv3576711 | Chr.9:133582804 - 133778331 on Build GRCh38 | Gain | VAV2 FAM163B DBH-AS1 SARDH DBH |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |