Assay Details
Target Gene Details
Entrez Gene ID: | 11202 |
Gene Name: | kallikrein related peptidase 8 |
Gene Aliases: |
HNP, NP, NRPN, PRSS19, TADG14 |
Location: |
Chr.19:50996008-51001702 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLK8 | NM_001281431.1 | NP_001268360.1 | ||
NM_007196.3 | NP_009127.1 | |||
NM_144505.2 | 3 | 310 | NP_653088.1 | |
NM_144506.2 | NP_653089.1 | |||
NM_144507.2 | NP_653090.1 | |||
NR_104008.1 | ||||
AA320217.1 | ||||
AB008390.2 | BAA82665.1 | |||
AB008927.1 | 2 | 152 | BAA82666.1 | |
AB009849.1 | BAA28673.1 | |||
AF095742.1 | AAD25979.1 | |||
AY359036.1 | AAQ89395.1 | |||
BM845575.1 | ||||
EU350538.1 | ||||
EU350539.1 | 3 | 212 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3644662 | Chr.19:50996542 - 51001686 on Build GRCh38 | Loss | KLK8 |
nsv1058277 | Chr.19:50985288 - 51006591 on Build GRCh38 | Gain | KLK8 KLK9 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |