Assay Details
Target Gene Details
Entrez Gene ID: | 80194 |
Gene Name: | transmembrane protein 134 |
Gene Aliases: |
- |
Location: |
Chr.11:67464348-67469277 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM134 | NM_001078650.2 | NP_001072118.1 | ||
NM_001078651.2 | NP_001072119.1 | |||
NM_025124.3 | NP_079400.1 | |||
NR_073409.1 | ||||
NR_073410.1 | ||||
NR_073411.1 | ||||
NR_073412.1 | ||||
NR_073413.1 | ||||
XM_006718693.2 | XP_006718756.1 | |||
XM_011545268.2 | XP_011543570.1 | |||
XM_017018356.1 | XP_016873845.1 | |||
AK025402.1 | BAB15126.1 | |||
AK296988.1 | ||||
AK303342.1 | ||||
AK310743.1 | ||||
AY007143.1 | ||||
BC125134.1 | ||||
BG488634.1 | ||||
BG719036.1 | ||||
BX400047.2 | ||||
CN336445.1 | ||||
DN999439.1 | ||||
HY053672.1 | ||||
HY059123.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv468604 | Chr.11:67418529 - 67516420 on Build GRCh38 | Loss | CARNS1 PITPNM1 PTPRCAP TMEM134 CORO1B GPR152 RPS6KB2 AIP CDK2AP2 MIR6752 CABP4 |
esv2422195 | Chr.11:67466851 - 67859430 on Build GRCh38 | Duplication | DOC2GP CABP2 PITPNM1 NDUFV1 TBX10 AIP GSTP1 ACY3 CDK2AP2 ALDH3B2 NUDT8 TMEM134 C11orf72 FAM86C2P MIR6752 |
dgv1977n54 | Chr.11:67397024 - 67521123 on Build GRCh38 | Loss | CARNS1 CABP2 PITPNM1 PTPRCAP AIP RAD9A CDK2AP2 CABP4 TMEM134 CORO1B GPR152 RPS6KB2 PPP1CA TBC1D10C MIR6752 |
dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion | CARNS1 CABP2 PITPNM1 PTPRCAP AIP RAD9A CDK2AP2 CABP4 LOC100130987 TMEM134 CORO1B GPR152 RPS6KB2 CLCF1 PPP1CA TBC1D10C MIR6752 |
nsv521723 | Chr.11:67408221 - 67543787 on Build GRCh38 | Loss | CARNS1 CABP2 PITPNM1 PTPRCAP AIP CDK2AP2 CABP4 TMEM134 CORO1B GPR152 RPS6KB2 TBC1D10C MIR6752 |
nsv527056 | Chr.11:67439955 - 67526837 on Build GRCh38 | Loss | CABP2 PITPNM1 TMEM134 CORO1B GPR152 AIP CDK2AP2 MIR6752 CABP4 |